Skip to Content
Merck
All Photos(1)

Key Documents

PHR1398

Supelco

Betamethasone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Betamethasone, 9α-Fluoro-11β,17α,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16β-methylprednisolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

certified reference material
pharmaceutical secondary standard

Agency

BP
EP
USP
traceable to BP 575
traceable to Ph. Eur. B1000000
traceable to USP 1066009

vapor pressure

<0.0000001 kPa ( 25 °C)

API family

betamethasone

CofA

current certificate can be downloaded

packaging

pkg of 1 g

storage condition

protect from light (20 mm aluminium crimp seal for unused portion)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to light yellow

mp

235-237 °C (lit.)

solubility

acetone: sparingly soluble
chloroform: very slightly soluble
ethanol: sparingly soluble
ether: very slightly soluble
methanol: sparingly soluble
water: insoluble

density

0.305 g/cm3 at 25 °C (77°F)

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-30°C

SMILES string

[H][C@@]12C[C@H](C)[C@](O)(C(=O)CO)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1

InChI key

UREBDLICKHMUKA-DVTGEIKXSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Betamethasone belongs to the corticosteriod family of active pharmaceutical ingredients (APIs).[1] It exhibits anti-inflammatory activity[1] and hence is used in the manufacture of various finished pharmaceutical products and is also employed as a starting material to manufacture other APIs that are related to this steroid family.[2]

Application

Betamethasone may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations[3][4] by using micellar liquid chromatography technique[3] and reversed-phase high-performance liquid chromatography technique.[4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA2174 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of dexamethasone and betamethasone in pharmaceuticals by reversed-phase HPLC
Pena-Garcia-Brioles D, et al.
Chromatographia, 39(9), 539-542 (1994)
Development and validation of a stability-indicating RP-HPLC method to separate low levels of dexamethasone and other related compounds from betamethasone
Xiong Y, et al.
Journal of Pharmaceutical and Biomedical Analysis, 49(3), 646-654 (2009)
Method development for betamethasone and dexamethasone by micellar liquid chromatography using cetyl trimethyl ammonium bromide and validation in tablets: Application to cocktails
Pena-Garcia-Brioles D, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(1), 65-71 (2004)
Development and validation of a stability-indicating RP-HPLC method for assay of betamethasone and estimation of its related compounds
Fu Q, et al.
Journal of Pharmaceutical and Biomedical Analysis, 51(3), 617-625 (2010)
M Gooderham et al.
The British journal of dermatology, 171(6), 1470-1477 (2014-07-02)
Plaque psoriasis has a relatively high prevalence in adolescence, resulting in a significant impact on quality of life, including social interactions. The primary objective was to assess the safety of once-daily application of fixed-combination calcipotriol plus betamethasone dipropionate gel in

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service