Skip to Content
Merck
All Photos(1)

Key Documents

B1055000

Betamethasone 21-valerate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37FO6
CAS Number:
Molecular Weight:
476.58
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(=O)OCC(=O)[C@@]1(O)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

InChI

1S/C27H37FO6/c1-5-6-7-23(32)34-15-22(31)27(33)16(2)12-20-19-9-8-17-13-18(29)10-11-24(17,3)26(19,28)21(30)14-25(20,27)4/h10-11,13,16,19-21,30,33H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1

InChI key

FEROCCAEIIKMJT-SUYDQAKGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone 21-valerate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of topical drugs within the skin, in particular glucocorticoids.
K Kubota et al.
Current problems in dermatology, 21, 61-66 (1993-01-01)
F Henry et al.
Skin pharmacology and applied skin physiology, 12(4), 199-210 (1999-07-27)
To assess the effect of the position of a limb (dependency) on the cutaneous vasoconstrictor (blanching) assay (VCA) using topical corticosteroid preparations. Two studies were performed on the forearms of healthy volunteers using tristimulus reflectance colorimetric assessments. The first one
Constantinos M Paleos et al.
Biomacromolecules, 5(2), 524-529 (2004-03-09)
A novel dendrimeric compound is designed with the objective of simultaneously addressing issues commonly encountered in drug delivery, i.e., stability in biological milieu as well as targeting. For this purpose, a multifunctional dendrimeric system derived from diaminobutane poly(propylene imine) dendrimers
S Tabolli et al.
Clinical and experimental dermatology, 34(3), 304-308 (2009-01-30)
The therapeutic effects of some histological staining agents on psoriasis have been reported in the past. One of these agents is eosin, which is still currently used in routine treatment, although it has never been formally tested in a randomized
K Kubota et al.
Dermatology (Basel, Switzerland), 188(1), 13-17 (1994-01-01)
The metabolism of betamethasone 17-valerate was estimated using an artificial living skin equivalent (LSE). Betamethasone 17-valerate, betamethasone 21-valerate and betamethasone were measured by a normal-phase high-performance liquid chromatographic (HPLC) method. Betamethasone 17-valerate was added to the culture medium with or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service