Skip to Content
Merck
All Photos(5)

Key Documents

T1502

Sigma-Aldrich

Triethanolamine hydrochloride

≥99.5% (titration)

Synonym(s):

2,2′,2′′-Nitrilotriethanol hydrochloride, Tris(2-hydroxyethyl)amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)3N · HCl
CAS Number:
Molecular Weight:
185.65
Beilstein:
3909940
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.5% (titration)

form

crystalline powder

useful pH range

7.3-8.3

pKa (25 °C)

7.8

mp

177-179 °C (lit.)

solubility

water: 0.333 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

Cl.OCCN(CCO)CCO

InChI

1S/C6H15NO3.ClH/c8-4-1-7(2-5-9)3-6-10;/h8-10H,1-6H2;1H

InChI key

HHLJUSLZGFYWKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethanolamine (TEA) belongs to the family of ethanolamine. It includes the properties of both amines and alcohols. Addition of TEA to an acidic buffer facilitates the determination of fluoride in inorganic orthophosphates by using a fluoride-selective electrode.

Application

Triethanolamine hydrochloride has been used:
  • in the preparation of triethanolamine buffer
  • in rinsing the digested sections, prepared for in situ hybridization
  • as a component of solution A to prepare TA-complement haemolysis buffer (TA-CHB)

Biochem/physiol Actions

Triethanolamine hydrochloride acts as a surfactant or pH adjuster. It is also used as an emulsifier in cosmetics and several topical medications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The serum complement system: a simplified laboratory exercise to measure the activity of an important component of the immune system
Inglis JE, et al.
Advances in Physiology Education, 32(4), 317-321 (2008)
Gender difference in hypothalamic-pituitary-adrenal axis response to alcohol in the rat: activational role of gonadal steroids
Ogilvie KM and Rivier C
Brain Research, 766(1-2), 19-28 (1997)
allergic Contact Dermatitis to Triethanolamine in a Child
Milanesi N, et al.
Pediatric Dermatology, 32(3), e112-e113 (2015)
Toxicology of mono-, di-, and triethanolamine
Reviews of Environmental Contamination and Toxicology, 1-86 (1997)
Optimal conditions for the kinetic assay of serum glutamate dehydrogenase activity at 37? C
Ellis G and Goldberg DM
Clinical Chemistry, 18(6), 523-527 (1972)

Protocols

Objective: To standardize a procedure for the enzymatic assay of Aprotinin.

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

See All

Related Content

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service