NIST2389A
Amino acids in 0.1 mol/L hydrochloric acid
NIST® SRM® 2389a
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 1.2 mL
manufacturer/tradename
NIST®
format
multi-component solution
Related Categories
General description
This Standard Reference Material (SRM) is intended primarily for use in calibration of chromatographic instrumentation for the determination of amino acids. SRM 2389a is a solution of 17 amino acids in a 0.1 mol/L aqueous solution of hydrochloric acid. For more information, please refer to the SDS and COA.
SRM 2389A_cert
SRM 2389A_SDS
SRM 2389A_cert
SRM 2389A_SDS
Application
- Alkaline sodium hypochlorite irrigant and its chemical interactions: Discusses the chemical interactions of sodium hypochlorite with other endodontic agents like EDTA and citric acid, emphasizing its role in tissue dissolution during dental procedures (Wright, Kahler, Walsh, 2017).
Other Notes
Certified for the analytes listed below.
Alanine (C3H7NO2), Arginine (C6H14N4O2), Aspartic acid (C4H7NO4), Glycine (C2H5NO2), Histidine (C6H9N3O2), Isoleucine (C6H13NO2), Leucine (C6H13NO2), Lysine (C6H14N2O2), Methionine (C5H11NO2S), Phenylalanine (C9H11NO2), Serine (C3H7NO3), Threonine (C4H9NO3), Tyrosine (C9H11NO3), Valine (C5H11NO2)
See certificate for values and more details at nist.gov/SRM.
Alanine (C3H7NO2), Arginine (C6H14N4O2), Aspartic acid (C4H7NO4), Glycine (C2H5NO2), Histidine (C6H9N3O2), Isoleucine (C6H13NO2), Leucine (C6H13NO2), Lysine (C6H14N2O2), Methionine (C5H11NO2S), Phenylalanine (C9H11NO2), Serine (C3H7NO3), Threonine (C4H9NO3), Tyrosine (C9H11NO3), Valine (C5H11NO2)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service