Skip to Content
Merck
All Photos(1)

Key Documents

LAA21

Supelco

L-Amino acids

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

Application

L-Amino acids has been used as a standard in the estimation of free amino acids of Centaurea. Solstitialis weed samples using reverse phase high performance liquid chromatography (RP-HPLC).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

21 L-Amino Acids plus glycine, 1 g of each:
  • A7627 L-Alanine
  • A5131 L-Arginine Hydrochloride
  • A0884 L-Asparagine
  • A9256 L-Aspartic Acid
  • C1276 L-Cysteine hydrochloride
  • C8755 L-Cystine
  • G1251 L-Glutamic Acid
  • G3126 L-Glutamine
  • G7126 Glycine
  • H8125 L-Histidine hydrochloride
  • H5534 trans-4-Hydroxy-L-proline
  • I2752 L-Isoleucine
  • L8000 L-Leucine
  • L5626 L-Lysine hydrochloride
  • M9625 L-Methionine
  • P2126 L-Phenylalanine
  • P0380 L-Proline
  • S4500 L-Serine
  • T8625 L-Threonine
  • T0254 L-Tryptophan
  • T3754 L-Tyrosine
  • V0500 L-Valine

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Olga A Sofyanovich et al.
PloS one, 14(5), e0216622-e0216622 (2019-05-10)
The role of glutathione (GSH) in eukaryotic cells is well known. The biosynthesis of this γ-glutamine tripeptide is well studied. However, other γ-glutamyl peptides were found in various sources, and the pathways of their formation were not always clear. The
Tomáš Pazderka et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 185, 207-216 (2017-06-04)
The Raman spectra of 20 proteinogenic amino acids were recorded in the solution, glass phase (as drop coating deposition Raman (DCDR) samples) and crystalline forms in the wide spectral range of 200-3200cm
HPLC determination of free nitrogenous compounds of Centaurea solstitialis (Asteraceae), the cause of equine nigropallidal encephalomalacia
Moret S, et al.
Toxicon, 46(6), 651-657 (2005)
Sílvia M F Bessada et al.
Journal of toxicology and environmental health. Part A, 80(13-15), 641-650 (2017-05-20)
Coleostephus myconis (L.) Rchb.f. (Asteraceae) is a highly disseminated plant species with ruderal and persistent growth. Owing to its advantageous agronomic properties, C. myconis might have industrial applications. However, this species needs to be comprehensively characterized before any potential use.
Aleksej Zelezniak et al.
Cell systems, 7(3), 269-283 (2018-09-10)
A challenge in solving the genotype-to-phenotype relationship is to predict a cell's metabolome, believed to correlate poorly with gene expression. Using comparative quantitative proteomics, we found that differential protein expression in 97 Saccharomyces cerevisiae kinase deletion strains is non-redundant and

Articles

Consider sequence length, solubility, and stability when designing peptides for custom synthesis.

Consider sequence length, solubility, and stability when designing peptides for custom synthesis.

Consider sequence length, solubility, and stability when designing peptides for custom synthesis.

Consider sequence length, solubility, and stability when designing peptides for custom synthesis.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service