Skip to Content
Merck

Skip To

A6282

Amino acid standards, physiological

analytical standard, basics

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

color

colorless

application(s)

food and beverages

format

neat

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Amino acids and related compounds are 2.5 μmoles per mL.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6407LAA10A2161
description

analytical standard, basics

description

analytical standard, acidics and neutrals

description

analytical standard

description

analytical standard

format

neat

format

neat

format

neat

format

multi-component solution

technique(s)

gas chromatography (GC): suitable, HPLC: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

color

colorless

color

colorless

color

-

color

-

analyte chemical class(es)

amino acids, peptides, proteins

analyte chemical class(es)

amino acids, peptides, proteins

analyte chemical class(es)

amino acids, peptides, proteins

analyte chemical class(es)

amino acids, peptides, proteins


Still not finding the right product?


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How long is the stability of this product once opened?

    1 answer
    1. This amino acid standard solution is fairly stable. The product is not assigned an expiration or retest date, however, a warranty period of 1 year from the date of shipment is offered. The stability of the product will be maintained throughout the warranty period provided that the product is stored and handled appropriately. Factors such as freeze/thaw, evaporation, and microbial or cross-contamination will impact the overall stability of the preparation.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review