Skip to Content
Merck
All Photos(3)

Key Documents

A5054

Sigma-Aldrich

Select agar

Synonym(s):

Agar, Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
50405901
NACRES:
NA.75

form

powder

Quality Level

application(s)

food and beverages

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Noble agar, also known as agar or agar-agar, is a gelling hydrocolloid extracted from sea weeds such as Gelidium and Gracilaria. It′s structure contains repeating units of D-galactose and 3,6-anhydro-L-galactose, with a few differences, in addition to a low ester sulphate residue. Agarose and agaropectin are the key components of agar. Noble agar is majorly used as both human and animal food supplement. In addition, it is also used in industries for the preparation of solid microbiological media.
Select agar is majorly used as a component of culture media for molecular genetics related studies.

Application

For use as a component of culture media for molecular genetics.
Select agar has been used as a gelling agent to prepare dielectric phantoms along with polyvinylpyrrolidone (PVP) and salts. It has also been used as a component of LB agar for the bacterial transformation screening.
Select agar has been used in the preparation of Luria Bertani (LB) agar plates during DNA transformation process.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Carlotta Ianniello et al.
Magnetic resonance in medicine, 80(1), 413-419 (2017-11-22)
To explore the use of polyvinylpyrrolidone (PVP) for simulated materials with tissue-equivalent dielectric properties. PVP and salt were used to control, respectively, relative permittivity and electrical conductivity in a collection of 63 samples with a range of solute concentrations. Their
Agar and agarose biotechnological applications
Armisen R
Hydrobiologia, 29(4), 157-166 (1991)
Construction and Cloning of Minigenes for in vivo Analysis of Potential Splice Mutations
Riedmayr LM, et al.
Bio-protocol, 8(4) (2018)
Agar
Food Gels, 1-51 (1990)
Structure of the agarose constituent of agar-agar
Araki C, et al.
Bulletin of the Chemical Society of Japan, 29(4), 543-544 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service