Skip to Content
Merck
All Photos(3)

Key Documents

A5431

Sigma-Aldrich

Noble agar

Synonym(s):

Agar, Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
50405901
NACRES:
NA.75

form

powder

Quality Level

application(s)

food and beverages

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Noble agar, also known as agar or agar-agar, is a gelling hydrocolloid extracted from sea weeds such as Gelidium and Gracilaria. It′s structure contains repeating units of D-galactose and 3,6-anhydro-L-galactose, with a few differences, in addition to a low ester sulphate residue. Agarose and agaropectin are the key components of agar. Noble agar is majorly used as both human and animal food supplement. In addition, it is also used in industries for the preparation of solid microbiological media.

Application

Noble agar has been used as a component of growth medium for High grade serous ovarian cancer (HGSOC) cell lines for soft agar colony assay. It has also been used as a media component to culture anaerobic bacteria, Bacteroides thetaitaomicron
Noble agar has been used to make agar gel plates that acts as a standard solid support material:
  • to culture tumor spheroids
  • in lysoplate assay method of Osserman & Lawlor to measure lysozyme concentration in albumen or plasma
  • as BG11 media component to culture Scenedesmus obtusiusculus strains
Originally designed for use in Noble′s cyanide citrate agar.

Preparation Note

Further purified from bacteriological agar with careful specific washings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 6

1 of 6

Agar and agarose biotechnological applications
Armisen R
Hydrobiologia, 157-166 (1991)
Effect of restricted preen-gland access on maternal self maintenance and reproductive investment in mallards
Giraudeau M, et al.
PLoS ONE, 5(10), e13555-e13555 (2010)
Characterization of Scenedesmus obtusiusculus AT-UAM for high-energy molecules accumulation: deeper insight into biotechnological potential of strains of the same species
Toledo-Cervantes A, et al.
Biotechnology reports (Amsterdam, Netherlands), 17, 16-23 (2018)
Maternal investment of female mallards is influenced by male carotenoid-based coloration
Giraudeau M, et al.
Proceedings of the Royal Society of London. Series B, Containing Papers of a Biological Character. Royal Society (Great Britain), 278(1706), 781-788 (2010)
Autophagy correlates with the therapeutic responsiveness of malignant pleural mesothelioma in 3D models
Barbone D, et al.
PLoS ONE, 10(8), e0134825-e0134825 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service