Skip to Content

A6686

Agar

Bacteriological, microbiology tested, suitable for plant cell culture, granular

Synonym(s):

Agar-agar, Gum agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
UNSPSC Code:
50405901
eCl@ss:
42040102
EC Number:
232-658-1
NACRES:
NA.28
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (Rhodophyceae)

Quality Segment

form

granular

technique(s)

cell culture | plant: suitable

mp

83-89  °C

transition temp

transition temp 32-39 °C

suitability

microbiology tested

application(s)

agriculture

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

General description

Agar, also known as agar-agar, is one of the biochemical agents most used in microbiology field. It is obtained from red algae Rhodophyceae, is the most abundantly used gelling agent in culture media. Agar is polysaccharide complex, composed of 70% agarose and 30% agaropectin.

Application

Agar has been used:
  • to culture Staphylococcus aureus for phage infection studies
  • as a supplement with NZCYM media for culturing Campylobacter coli strains for lysogeny screening
  • as a basic media component to improve in vitro ovule and embryo culture methods of cross Chitalpa hybrids to facilitate the rescue of triploids
  • in the culture media for Bdellovibrio spp. and Micavibrio spp. strains to check their susceptibility towards a variety of select agents

Other Notes

A purified agar from which the naturally occurring pigments, salts and miscellaneous matter has been reduced to a minimum.


Still not finding the right product?

Explore all of our products under Agar


Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library