Skip to Content
Merck
All Photos(1)

Key Documents

H-100

Supelco

17α-Hydroxyprogesterone-2,3,4-13C3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C18H30O3
CAS Number:
Molecular Weight:
333.44
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)C1(O)CCC2C3CCC4=CC(=O)CCC4(C)C3CCC12C

InChI

1S/C21H30O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12,16-18,24H,4-11H2,1-3H3/t16-,17+,18+,19+,20+,21+/m1/s1/i6+1,12+1,15+1

InChI key

DBPWSSGDRRHUNT-GGSYSODVSA-N

General description

A stable labeled internal standard suitable for quantitation of 17α-Hydroxyprogesterone levels in LC-MS/MS testing assays for newborn screening or endocrinology. For LC-MS/MS assays 13C labeled internal standards can offer certain advantages such as eliminating potential issues of isotopic exchange and minimizing separation of later-eluting analytes from a higher substituted deuterium-labeled internal standard.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Peng et al.
Journal of immunology (Baltimore, Md. : 1950), 183(10), 6058-6068 (2009-10-30)
Regulation of T cell immunity by C5a has been suggested from recent studies. However, the underlying mechanisms, particularly the involved cells and biochemical basis, are not well defined. In this study, the direct modulation of dendritic cell (DC) activation and
Lifeng Tian et al.
Oncotarget, 5(17), 7303-7315 (2014-09-18)
In our prior publications we characterized a conserved acetylation motif (K(R)xxKK) of evolutionarily related nuclear receptors. Recent reports showed that peroxisome proliferator activated receptor gamma (PPARγ) deacetylation by SIRT1 is involved in delaying cellular senescence and maintaining the brown remodeling
O Rosendo et al.
Journal of dairy science, 87(8), 2535-2545 (2004-08-26)
Fifty-two multiparous Holstein cows were randomly assigned to receive 0 or 20 mg of biotin/d starting at an average of 16 d prepartum and then switched to 0 or 30 mg of biotin/d from calving through 70 d postpartum to
Tim Hebbrecht et al.
PloS one, 12(9), e0185076-e0185076 (2017-09-25)
Invasive cancer cells develop small actin-based protrusions called invadopodia, which perform a primordial role in metastasis and extracellular matrix remodelling. Neural Wiskott-Aldrich syndrome protein (N-WASp) is a scaffold protein which can directly bind to actin monomers and Arp2/3 and is
James Hulit et al.
Molecular and cellular biology, 24(17), 7598-7611 (2004-08-18)
Constitutive beta-catenin/Tcf activity, the primary transforming events in colorectal carcinoma, occurs through induction of the Wnt pathway or APC gene mutations that cause familial adenomatous polyposis. Mice carrying Apc mutations in their germ line (ApcMin) develop intestinal adenomas. Here, the

Articles

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service