Skip to Content
Merck
All Photos(1)

Key Documents

802905

Sigma-Aldrich

Corticosterone-9,11,12,12-d4

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D4H26O4
Molecular Weight:
350.49
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@]12C(CC[C@]3([H])[C@]2([2H])[C@@]([2H])(O)C([2H])([2H])[C@@]4(C)[C@@]3([H])CC[C@@H]4C(CO)=O)=CC(CC1)=O

InChI

1S/C21H30O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h9,14-17,19,22,24H,3-8,10-11H2,1-2H3/t14-,15-,16+,17-,19+,20-,21-/m0/s1/i10D2,17D,19D

InChI key

OMFXVFTZEKFJBZ-HECNVQIESA-N

Related Categories

Application

This product may be used as an analytical standard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Sze et al.
Journal of neuroendocrinology, 33(1), e12916-e12916 (2020-12-04)
Acute swim stress results in the robust production of several neuroactive steroids, which act as mediators of the stress response. These steroids include glucocorticoids, and positive GABAA receptor modulatory steroids such as allopregnanolone and tetrahydrocorticosterone (THDOC), which potentiate inhibitory GABA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service