Skip to Content
Merck
All Photos(1)

Documents

C-106

Supelco

Cortisol solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Hydrocortisone solution, 17-Hydroxycorticosterone, Cortisol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Cortisol or hydrocortisone, also known as the stress hormone, is aglucocorticoid steroid. It is released by the adrenal glands and is involved inmaintaining the circadian rhythm. The hormone is measured in whole blood by LC-MS/MS for newborn screening of congenital adrenal hyperplasia (CAH) and in serum or plasma by LC-MS/MS for determination of several diseases including Cushing’s syndrome, Addison’s disease, and Sheehan’s syndrome.

Application

This Certified Reference Material (CRM) in solution can also be used as follows:
  • Development of a disposable immunosensor based on direct competitive-enzyme linked immunoassay (DC-ELISA) to determine cortisol in human saliva samples
  • Quantitative analysis of cortisol in human hair samples by a liquid-chromatography-tandem mass spectrometric (LC-MS/MS) method
  • Selective and sensitive analysis of both cortisol and cortisone in human hair samples by an automated solid phase extraction (SPE) method combined with liquid chromatography (LC) coupled to electrospray ionization-tandem mass spectrometry (ESI-MS3)
  • Multi-residue determination of cortisol, cortisone, and their three tetrahydro metabolites from human urine samples by an LC-MS/MS method
  • Human saliva analysis for the estimation of cortisol and cortisone by ionic liquid dispersive liquid-liquid microextraction (IL-DLLME) and liquid chromatography combined with ultra-violet/visible (UV/Vis) detection

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanda Tesanović et al.
Collegium antropologicum, 37(2), 367-371 (2013-08-15)
We conducted this study to determine the degree of obesity influence on the hypoglycemic response of growth hormone and cortisol after weight loss of 5%. A total of 45 non-diabetic, male subjects followed in the outpatient endocrinological departments were divided
M V Bezruchko et al.
Klinichna khirurhiia, (3)(3), 22-25 (2013-05-31)
The results of comparison between the operation stress degree in various kinds of surgical interventions, performed for an acute cholecystitis, using determination of cortizol, prolactin and glucose content before the operation, intraoperatively and postoperatively in 50 patients, are adduced. There
Miriam Diane Lense et al.
American journal on intellectual and developmental disabilities, 118(3), 201-210 (2013-06-06)
Williams syndrome (WS) is a neurodevelopmental genetic disorder associated with high rates of anxiety and social issues. We examined diurnal cortisol, a biomarker of the stress response, in adults with WS in novel and familiar settings, and compared these profiles
Benjamin P Casavant et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(25), 10111-10116 (2013-06-05)
Although the field of microfluidics has made significant progress in bringing new tools to address biological questions, the accessibility and adoption of microfluidics within the life sciences are still limited. Open microfluidic systems have the potential to lower the barriers
Wanpen Turakitwanakan et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 96 Suppl 1, S90-S95 (2013-06-04)
Mindfulness meditation is a method to relax the mind that decreases stress, which otherwise would increase serum cortisol. So, mindfulness meditation should decrease serum cortisol. To study the effect of mindfulness meditation on mental health by using Thai GHQ28 questionaire

Articles

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service