Merck
  • Preliminary report of hypoglycemic response in obese metabolic syndrome males treated with metformin after weight loss intervention.
Preliminary report of hypoglycemic response in obese metabolic syndrome males treated with metformin after weight loss intervention.
Collegium antropologicum (2013-08-15)
Sanda Tesanović, Maja Radman, Desa Tesanović, Dubravka Jurisić Erzen, Izet Hozo
ABSTRACT
We conducted this study to determine the degree of obesity influence on the hypoglycemic response of growth hormone and cortisol after weight loss of 5%. A total of 45 non-diabetic, male subjects followed in the outpatient endocrinological departments were divided into three groups comprising 15 subjects in each group, based upon body mass index (BMI) to healthy, overweight and obese group. Metformin was administered in the dose of 50 mg daily to the overweight and obese participants. Cortisol was measured at 0, 60 and 120 minutes. Growth hormone (GH) was measured at -15, 0, 30, 60, 90 and 120 minutes. Values of cortisol and GH were compared upon changes in hypothalamo-pituitary-adrenal (HPA) response to insulin induced hypoglycemia initially and after weight loss of 5% for overweight and obese participants. The BMI of the healthy group ranged 20.0-24.5 kg/m2 (median: 22.8); overweight group ranged 25.9-29.7 kg/m2 (median: 28.3); and obese group ranged 30.9-34.6 kg/m2 (median: 32.6). There were no significant differences of cortisol values among groups at 0 (chi2 = 2.0; p = 0.365); 60 (chi2 = 0.754; P = 0.686) and at 120 minutes (chi2 = 0.466; p = 0.792). The comparisons among groups were significant for differences of GH values at -15 (chi2 = 25.0; p < 0.01); 0 (chi2 = 16.2; p < 0.01); 30 (chi2 = 16.2; p < 0.01); 60 (chi2 = 32.8; p < 0.01); 90 (chi2 = 30.2; p < 0.01) and at 120 minutes (chi2 = 27.3; p < 0.01). Healthy and obese subjects significantly differed in growth hormone response at -15 (Z = 4.67; p < 0.01); 0 (Z = 3.83; p < 0.01); 60 (Z = 2.78; p = 0.05); 90 (Z = 4.67; p < 0.01) and at 120 minutes (Z = 4.23; p < 0.01). Changes on the various levels of HPA axis, when it is activated by a stress as it is the case in insulin-induced hypoglycemia correspond to the degree of obesity. Weight loss of 5% was not enough for restoration of a normal stimulated growth hormone release and did not influence on the level of cortisol.
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture