Merck
  • Diurnal cortisol profile in Williams syndrome in novel and familiar settings.
Diurnal cortisol profile in Williams syndrome in novel and familiar settings.
American journal on intellectual and developmental disabilities (2013-06-06)
Miriam Diane Lense, Andrew J Tomarken, Elisabeth M Dykens
ABSTRACT
Williams syndrome (WS) is a neurodevelopmental genetic disorder associated with high rates of anxiety and social issues. We examined diurnal cortisol, a biomarker of the stress response, in adults with WS in novel and familiar settings, and compared these profiles to typically developing (TD) adults. WS and TD participants had similar profiles in a familiar setting, while participants with WS had elevated cortisol late in the day in the novel setting when social demands were higher. The cortisol awakening response in WS was associated with parent-reported levels of somatic complaints and social difficulties. Results suggest that adults with WS have a typical diurnal cortisol profile that may be sensitive to social and activity transitions throughout the day.
Product Description
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Pricing and availability is not currently available.