Skip to Content
Merck
All Photos(1)

Key Documents

483702

Sigma-Aldrich

Germanium(IV) oxide

≥99.99% trace metals basis

Synonym(s):

Germanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GeO2
CAS Number:
Molecular Weight:
104.64
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

powder

mp

>400 °C (lit.)

solubility

ethylene glycol: soluble

density

4.23 g/cm3

application(s)

battery manufacturing

SMILES string

O=[Ge]=O

InChI

1S/GeO2/c2-1-3

InChI key

YBMRDBCBODYGJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Germanium(VI) oxide, also known as germania or germanium dioxide, is a white or off-white powder. It is a refractory metal oxide with a high melting point (1,600 °C) and offers excellent chemical resistance. In addition, germanium(VI) oxide has excellent optoelectronic properties that have made it a valuable material in the semiconducting industry. It has a high refractive index of approximately 2.1, which is higher than that of many other semiconductor materials, and a low absorption coefficient. These properties make it useful for applications such as waveguides, antireflective coatings, and high-index contrast coatings. In addition, germanium(VI) has a high bandgap of about 3.1 eV coupled with relatively high transparency in the infrared region of the electromagnetic spectrum, which makes it useful for applications such as infrared detectors, LEDs, and solar cells.

Application

Building block for inorganic nanowires. Used in the synthesis of ultra-high refractive index and ultra-low loss optical waveguides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Liang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(71), 17879-17884 (2017-11-10)
The attractive properties of layered inorganic materials, which make them suitable for numerous applications in chemical industries and life sciences, originated from their crystalline framework structures. Here, we report a new layered germanate PKU-21, which was prepared by the hydrothermal
Rao, C.N.R., et al.
Journal of Materials Chemistry, 14, 440-440 (2004)
Imoto, K.
Optronics, 260, 116-116 (2003)
V V Dvoyrin et al.
Optics express, 19(9), 7992-7999 (2011-06-07)
A novel all-fiber laser based on a highly GeO2-doped dispersion-shifted Tm-codoped fiber, pumped at 1.56 µm wavelength and lasing at 1.862 µm wavelength with a slope efficiency up to 37% was demonstrated. The single-mode Tm-doped fiber with the 55GeO2-45SiO2 core
Xinguo Hong et al.
The Review of scientific instruments, 80(7), 073908-073908 (2009-08-07)
We describe an approach for acquiring high quality x-ray absorption fine structure (XAFS) spectroscopy spectra with wide energy range at high pressure using diamond anvil cell (DAC). Overcoming the serious interference of diamond Bragg peaks is essential for combining XAFS

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service