Skip to Content
Merck
All Photos(1)

Documents

396362

Sigma-Aldrich

Germanium(IV) methoxide

Synonym(s):

Methyl germanate(IV), Tetramethoxygermanium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ge(OCH3)4
CAS Number:
Molecular Weight:
196.78
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

core: germanium
reagent type: catalyst

impurities

≤5% methyl alcohol

refractive index

n20/D 1.399 (lit.)

bp

66-67 °C/36 mmHg (lit.)

mp

−18 °C (dec.) (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

CO[Ge](OC)(OC)OC

InChI

1S/C4H12GeO4/c1-6-5(7-2,8-3)9-4/h1-4H3

InChI key

ACOVYJCRYLWRLR-UHFFFAOYSA-N

Application

  • Used as a promoting agent for vanadyl pyrophosphate-catalyzed oxidation reactions
  • Used to prepare sol-gel hybrid organic-inorganic coatings for capillary microextension and gas chromatography (GC)
  • For preparation of metal(IV) β-cyclodextrin bucket wheel complex
  • Preparation of gold nanoparticle templated germania nanoshells
Sol-gel intermediate in the formation of GeS2 and IR-transparent optics.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 2 - STOT SE 3

Target Organs

Eyes, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F

Flash Point(C)

41 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceram. Bull., 63, 1427-1427 (1984)
Journal of the American Chemical Society, 112, 5636-5636 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service