Skip to Content
Merck
All Photos(2)

Documents

483001

Sigma-Aldrich

Germanium(IV) oxide

powder, 99.999% trace metals basis

Synonym(s):

Germanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GeO2
CAS Number:
Molecular Weight:
104.64
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

powder

mp

>400 °C (lit.)

density

4.23 g/cm3

application(s)

battery manufacturing

SMILES string

O=[Ge]=O

InChI

1S/GeO2/c2-1-3

InChI key

YBMRDBCBODYGJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xitong Chen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(49), 11913-11919 (2017-08-12)
The development of novel photocatalysts usually centers on features such as band structures, various nano-, micro-, or macro-forms, and composites in efforts to tune their light absorption and charge separation efficiency. In comparison, the selectivity of photocatalysts with respect to
E A Anashkina et al.
Optics express, 20(24), 27102-27107 (2012-11-29)
We report generation of femtosecond optical pulses tunable in the 1.6-2.5 μm range using GeO2-doped core silica-cladding fibers. Optical solitons with a duration of 80-160 fs have been measured by the FROG technique in the 2-2.3 μm range. To the
Xin-ben Zhang et al.
Optics express, 20(18), 19799-19805 (2012-10-06)
A GeO2 doped triangular-core photonic-crystal fiber (PCF) is designed and fabricated to allow the generation of a hollow beam through a nonlinear-optical transformation by femtosecond pulses at 1040 nm from a high power Yb-doped PCF laser oscillator. The hollow beam
Quintin Cheek et al.
ACS nano, 14(3), 2869-2879 (2020-02-23)
The growth of Ge nanowires in water inside a liquid transmission electron microscope (TEM) holder has been demonstrated at room temperature. Each nanowire growth event was stimulated by the incident electron beam on otherwise unsupported liquid Ga or liquid In
Sarah L Sewell et al.
Dalton transactions (Cambridge, England : 2003), (29)(29), 3857-3865 (2008-07-17)
Biomimetic synthesis is emerging as an advantageous alternative to the harsh synthetic conditions traditionally used in metal oxide syntheses techniques. Silaffins, proteins from the C. fusiformis diatom, form silica in an aqueous environment under benign conditions. Amine terminated PAMAM and

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service