Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Germanium(IV) oxide
Synonym(s):
Germanium dioxide, Germanium(IV) oxide
Linear Formula:
GeO
2
CAS Number:
1310-53-8
Molecular Weight:
104.64
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
483702
Germanium(IV) oxide,
≥99.99% trace metals basis
Expand
View Pricing
199478
Germanium(IV) oxide,
(crystalline powder), 99.998% trace metals basis
Expand
View Pricing
483001
Germanium(IV) oxide,
powder, 99.999% trace metals basis
Expand
View Pricing
1.12177
Germanium(IV) oxide,
99+
Expand
View Pricing