Skip to Content
Merck
All Photos(1)

Key Documents

310670

Sigma-Aldrich

Oxalyl chloride solution

2.0 M in methylene chloride

Synonym(s):

Dichlorooxalic acid, Ethanedioyl chloride, Oxalic acid chloride, Oxalic acid dichloride, Oxalic dichloride, Oxaloyl chloride, Oxaloyl dichloride, Oxalyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCOCl
CAS Number:
Molecular Weight:
126.93
Beilstein:
1361988
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: oxidant

concentration

2.0 M in methylene chloride

density

1.335 g/mL at 25 °C

functional group

acyl chloride

SMILES string

ClC(=O)C(Cl)=O

InChI

1S/C2Cl2O2/c3-1(5)2(4)6

InChI key

CTSLXHKWHWQRSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxalyl chloride is generally used as a dimethyl sulfoxide activator and a chlorinating agent to convert carboxylic acid to acid chlorides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of highly soluble and heat stable new poly (amide-ether) s containing pyridine rings in the main chain.
Banihashemi A and Vakili MR
e-Polymers, 8(1) (2008)
Catalytic syntheses of N-heterocyclic ynones and ynediones by in situ activation of carboxylic acids with oxalyl chloride.
Christina Boersch et al.
Angewandte Chemie (International ed. in English), 50(44), 10448-10452 (2011-09-13)
Peter J Manley et al.
Organic letters, 4(18), 3127-3129 (2002-08-31)
[reaction: see text] A mild, practical, one-pot method for the generation of imidoyl chlorides and their subsequent in situ reaction with pyridine-1-oxides is described. The imidoyl chlorides were formed from the reaction of secondary amides with a stoichiometric amount of
Oxidation of long-chain and related alcohols to carbonyls by dimethyl sulfoxide" activated" by oxalyl chloride.
Mancuso AJ, et al.
The Journal of Organic Chemistry, 43(12), 2480-2482 (1978)
Benoît Heurtaux et al.
The Journal of organic chemistry, 70(4), 1474-1477 (2005-02-12)
[reaction: see text] Several natural pulvinic acids were synthesized. Silyl ketene acetals derived from methyl arylacetates (4 equiv) reacted with oxalyl chloride at -78 degrees C, without the need of adding a catalyst. After treatment of the crude diketones with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service