Skip to Content
Merck
All Photos(1)

Documents

320536

Sigma-Aldrich

Thionyl chloride

reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SOCl2
CAS Number:
Molecular Weight:
118.97
Beilstein:
1209273
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor pressure

97 mmHg ( 20 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

79 °C (lit.)

mp

−105 °C (lit.)

density

1.631 g/mL at 25 °C (lit.)

SMILES string

ClS(Cl)=O

InChI

1S/Cl2OS/c1-4(2)3

InChI key

FYSNRJHAOHDILO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thionyl chloride is an inorganic acid chloride mainly used as a reagent to prepare carboxylic acid chlorides from carboxylic acids.

Application

Thionyl chloride may be used in the following processes:
  • To facilitate the synthesis of indenones from 3-hydroxyindanones via dehydroxylation.
  • To functionalize silica gel for thioacetalization of aldehydes.
  • Conversion of tert-butyl esters to acid chlorides.
  • Conversion of aliphatic and aromatic sulfoxides to sulfides in the presence of triphenylphosphine.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Chemistry, 607-607 null
Deoxygenation of sulfoxides to sulfides with thionyl chloride and triphenylphosphine: Competition with the Pummerer reaction
Jang Y, et al.
The Journal of Organic Chemistry, 78(12), 6328-6331 (2013)
The Conversion of tert-Butyl Esters to Acid Chlorides Using Thionyl Chloride.
Greenberg JA & Sammakia T
The Journal of Organic Chemistry, 82(6), 3245-3251 (2017)
Selective protection of carbonyl compounds. Silica gel treated with thionyl chloride as an effective catalyst for thioacetalization.
Kamitori Y, et al.
The Journal of Organic Chemistry, 51(9), 1427-1431 (1986)
Thionyl chloride mediated dehydroxylation of 3-hydroxyindanones to indenones.
Chanda T, et al.
Tetrahedron Letters, 56(31), 4603-4606 (2015)

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service