Skip to Content
Merck
All Photos(4)

Key Documents

270997

Sigma-Aldrich

Dichloromethane

anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer

Synonym(s):

DCM, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
technique(s):
gas chromatography (GC): suitable
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
6.86 psi ( 20 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Agency

suitable for EPA 1613

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
6.86 psi ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

1223 °F

contains

40-150 ppm amylene as stabilizer

expl. lim.

22 %

technique(s)

gas chromatography (GC): suitable

impurities

≤0.001% water
≤0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane has been tested as a solvent medium for the dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml.[1] Role of quantity of TiO2 loading on activated carbon support employed in the photodecomposition of dichloromethane has been investigated.[2]
Dichloromethane is a chlorinated organic solvent that is susceptible to degradation with time. This can be suppressed by adding amylene as a stabilizer.

Application

Dichloromethane has been used for the quantification of herbivore (gypsy moth) induced blueberry volatiles by gas chromatography (GC).[3]
Dichloromethane may be used as a solvent for the oxidation of primary and secondary alcohols using dipyridine-chromium(VI) oxide.[1] Lithium perchlorate suspended in DCM is an effective medium for Mukaiyama aldol reactions of aldehydes with silyl ketene acetal.[4] DCM may also be used as solvent for the synthesis of 2-bromo-6-lithiopyridine from 2,6-dibromopyridine via lithium-bromine exchange reaction.[5]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cesar R Rodriguez-Saona
Journal of visualized experiments : JoVE, (58)(58), doi:10-doi:10 (2012-01-05)
Herbivore-induced plant volatiles (HIPVs) are commonly emitted from plants after herbivore attack. These HIPVs are mainly regulated by the defensive plant hormone jasmonic acid (JA) and its volatile derivative methyl jasmonate (MeJA). Over the past 3 decades researchers have documented
Carbon-carbon bond formation catalyzed by lithium perchlorate in dichloromethane
Reetz MT & Fox DN
Tetrahedron Letters, 34(7), 1119-1122 (1993)
Efficient Preparation of 2-Bromo-6-lithiopyridine via Lithium? Bromine Exchange in Dichloromethane
Peterson MA & Mitchell JR
The Journal of Organic Chemistry, 62(23), 8237-8239 (1997)
Dipyridine-chromium (VI) oxide oxidation of alcohols in dichloromethane.
Collins JC, et al.
Tetrahedron Letters, 9(30), 3363-3366 (1968)
Effect of activated carbon content in TiO2-loaded activated carbon on photodegradation behaviors of dichloromethane.
Torimoto T, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 103(1), 153-157 (1997)

Articles

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service