Skip to Content
Merck
All Photos(1)

Documents

280909

Sigma-Aldrich

(Chloromethylene)dimethyliminium chloride

95%

Synonym(s):

(Chlormethylene)dimethylammonium chloride, Arnold′s reagent, Dimethylchloroformiminium chloride, Vilsmeier reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=N(CH3)2Cl
CAS Number:
Molecular Weight:
128.00
Beilstein:
3566322
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

impurities

<5% DMF

mp

132 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Cl-].C\[N+](C)=C\Cl

InChI

1S/C3H7ClN.ClH/c1-5(2)3-4;/h3H,1-2H3;1H/q+1;/p-1

InChI key

QQVDYSUDFZZPSU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reagent or Reactant for the synthesis of:
  • β-Lactams vis the Staudinger reaction
  • N-vinyl substituted indoles via cross coupling of NH-indoles with vinyl bromides
  • Multi ring-fused 2-pyridone-based fluorescent scaffold
  • ester prodrugs as anti-poxvirus agents
  • Trimethine cyanine dyes for sensing G-quadruplex formation
  • Phospha-isosteres via Hundsdiecker-Barton iododecacarboxylation
  • 2-Azetidinones via [2+2]-cycloaddition

Used for studying organic photosensitizers for application in dye-sensitized solar cells
Reagent used to prepare ß-lactams from ß-amino acids via carboxyl activation and cyclodehydration. Also used to convert α-aminonitriles into 4-chloroimidazoles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 5451-5451 (2006)
J. Chem. Res. (M), 705-705 (2005)
Azhar Abbas et al.
Bioorganic chemistry, 82, 163-177 (2018-10-16)
Benzohydrazide derivatives 1-43 were synthesized via "one-pot" reaction and structural characterization of these synthetic derivatives was carried out by different spectroscopic techniques such as 1H NMR and EI-MS. The synthetic molecules were evaluated for their in vitro urease inhibitory activity.
Martin Mlateček et al.
Dalton transactions (Cambridge, England : 2003), 44(46), 20242-20253 (2015-11-06)
The reaction of PhBCl2 with 1H-1,2,4-λ(3)-diazaphosphole in the presence of NEt3 gives a new scorpionate ligand, phenyl-tris(1,2,4-diazaphospholyl)borate (PhTdap). The coordination behaviour of this ligand toward transition and non-transition metals has been comprehensively studied. In the thallium(I) complex, Tl(PhTdap), κ(2)-N,N bonding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service