Skip to Content
Merck
All Photos(2)

Documents

241474

Sigma-Aldrich

Silver sulfide

99.9% trace metals basis

Synonym(s):

Disilver sulfide, Silver(I) sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag2S
CAS Number:
Molecular Weight:
247.80
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

mp

845 °C (lit.)

density

7.234 g/mL at 25 °C (lit.)

SMILES string

[Ag]S[Ag]

InChI

1S/2Ag.S

InChI key

OCWNMDUAUUAHSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gururaj M Neelgund et al.
Colloids and surfaces. B, Biointerfaces, 100, 215-221 (2012-07-07)
Herein we report the design of antimicrobial nanohybrids, f-MWCNTs-CdS and f-MWCNTs-Ag(2)S developed by covalent grafting of cationic hyperbranched dendritic polyamidoamine (PAMAM) onto multiwalled carbon nanotubes (MWCNTs) and successive deposition of CdS and Ag(2)S quantum dots (QDs). The CdS and Ag(2)S
Jingyu Liu et al.
Environmental science & technology, 45(17), 7345-7353 (2011-07-21)
Among the many new engineered nanomaterials, nanosilver is one of the highest priority cases for environmental risk assessment. Recent analysis of field samples from water treatment facilities suggests that silver is converted to silver sulfide, whose very low solubility may
Yan Lei et al.
Journal of the American Chemical Society, 134(42), 17392-17395 (2012-10-10)
P3HT:Ag(2)S hybrid solar cells with broad absorption from the UV to NIR band were directly fabricated on ITO glass by using a room temperature, low energy consumption, and low-cost soft-chemical strategy. The resulting Ag(2)S nanosheet arrays facilitate the construction of
Jun Chen et al.
Nanoscale, 4(15), 4455-4458 (2012-06-28)
Herein, a novel structural transformation of Ag(2)S nanoparticles from hollow particles to solid spheres is reported. The features of these structures are identified through a set of characterizations based on which the formation mechanism is also investigated.
Zhongping Liu et al.
Nanoscale, 5(4), 1570-1575 (2013-01-19)
In this work, AgInS(2) hierarchical flowerlike nanoarchitectures, which are composed of ultrathin nanowires, were synthesized by thermolysis of a mixed solution of AgNO(3), InCl(3)·4H(2)O and n-dodecanethiol at elevated temperature. The average diameter and length of the nanowires composing the nanoarchitectures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service