All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Bi2S3
CAS Number:
Molecular Weight:
514.16
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Assay:
99%
form:
powder
Recommended Products
Quality Level
Assay
99%
form
powder
density
7.7 g/mL at 25 °C (lit.)
SMILES string
S=[Bi]S[Bi]=S
InChI
1S/2Bi.3S
InChI key
ZABYETDLLXARHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Bismuth(III) sulphide can be precipitated by hydrogen sulphide from bismuth solutions. 1
Packaging
Packaged in glass bottles
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
C J Tang et al.
Journal of nanoscience and nanotechnology, 10(8), 5451-5455 (2010-12-04)
The spherical Bi2S3 flowers have been fabricated by a facile environmentally friendly hydrothermal method. It was found that the flowers are composed of pure orthorhombic phase Bi2S3, the nanorods (nanowires) composed of the flowers grow radically from a center toward
Juan Lu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 29(1), 48-51 (2009-04-24)
In the present study, bismuth sulfide (Bi2S3) nanoribbons were prepared by the hydrothermal method using bismuth nitrate (Bi(NO3)3 x 5H2O), thioacetamide (C2H5NS) and nitrilotriacetic acid (C6H9NO6) as raw materials at 180 degrees C for 12 h. The reaction time was
Synthesis of a highly ordered single-crystalline Bi2S3 nanowire array and its metal/semiconductor/metal back-to-back Schottky diode.
Haifeng Bao et al.
Small (Weinheim an der Bergstrasse, Germany), 4(8), 1125-1129 (2008-07-25)
X-ray computed tomography imaging of breast cancer by using targeted peptide-labeled bismuth sulfide nanoparticles.
Joseph M Kinsella et al.
Angewandte Chemie (International ed. in English), 50(51), 12308-12311 (2011-10-27)
Solution-processed heterojunction solar cells based on p-type PbS quantum dots and n-type Bi2 S3 nanocrystals.
Arup K Rath et al.
Advanced materials (Deerfield Beach, Fla.), 23(32), 3712-3717 (2011-07-07)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service