Skip to Content
Merck
All Photos(2)

Key Documents

204404

Sigma-Aldrich

Silver iodide

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgI
CAS Number:
Molecular Weight:
234.77
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

Assay

99.999% trace metals basis

form

powder and chunks

impurities

≤20.0 ppm Trace Metal Analysis

density

5.68 g/mL at 25 °C (lit.)

SMILES string

[Ag]

InChI

1S/Ag.HI/h;1H/q+1;/p-1

InChI key

MSFPLIAKTHOCQP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chikako Kuwabara et al.
Cryobiology, 64(3), 279-285 (2012-03-13)
In this study, we examined the effects on freezing of 26 kinds of flavonoid compounds, which were randomly selected as compounds with structures similar to those of flavonoid compounds existing in deep supercooling xylem parenchyma cells (XPCs) in trees, in
Biao Xu et al.
Small (Weinheim an der Bergstrasse, Germany), 7(24), 3439-3444 (2011-11-01)
AgI quantum dot (QD)-based room-temperature fast ionic conductors are prepared via an aqueous route. New phase behavior and good performance are found in this material. This is a prototype of quantum dot-ionics.
Yu Sakurai et al.
The Tohoku journal of experimental medicine, 228(4), 317-323 (2012-11-08)
Nanomaterials have great potential in the field of medicine and have been studied extensively. In a previous study, we addressed the potential of silver iodide (AgI) as X-ray contrast media, because it possessed high imaging ability in the measurement by
Gertruida J S Venter et al.
Acta crystallographica. Section B, Structural science, 65(Pt 2), 182-188 (2009-03-21)
The reaction of silver(I) iodide with tri(p-tolyl)phosphine in MeCN solution in 1:3 molar ratio yields a polymorph of the complex of the formula [AgI{P(4-MeC6H4)3}3], with the Ag atom in a distorted tetrahedral environment. A polymorphic structure of this complex (a)
Xuexiang Hu et al.
Environmental science & technology, 44(18), 7058-7062 (2010-08-26)
The plasmon-induced photocatalytic inactivation of enteric pathogenic microorganisms in water using Ag-AgI/Al(2)O(3) under visible-light irradiation was investigated. The catalyst was found to be highly effective at killing Shigella dysenteriae (S. dysenteriae), Escherichia coli (E. coli), and human rotavirus type 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service