Skip to Content
Merck
All Photos(3)

Key Documents

342459

Sigma-Aldrich

Copper(I) sulfide

powder, −325 mesh

Synonym(s):

Cuprous sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu2S
CAS Number:
Molecular Weight:
159.16
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

particle size

−325 mesh

density

5.6 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Cu]S[Cu]

InChI

1S/2Cu.S

InChI key

JESPAFKOCOFQIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Copper sulfide shows metal-like electrical conductivity, chemical-sensing capability and suitable characteristics for absorption of solar energy. Cu63 NMR, X ray photoelectron spectroscopy,2 copper sulfide CuS was studied in detail. Copper sulfide is a monovalent and oxidation state was determined to be 2.2

Application


  • Pharmacological Study on Ocfentanil: Research on the pharmacological properties of Ocfentanil, including its biochemistry and potent analgesic effects, was explored in a study where toxicology and behavioral impacts on zebrafish larvae and mice were assessed, confirming its potential for pain management applications (Bilel et al., 2023).

  • Ocfentanil in Surgical Procedures: Ocfentanil′s role in surgical applications was highlighted in a study where new analytical methods were developed for its detection in biological samples, emphasizing its significance in pharmacological studies and surgical pain management (Jung et al., 2020).

  • Ocfentanil Solution for Pharmacological Studies: The development and validation of a method for detecting Ocfentanil in hair samples indicate its critical role in forensic and pharmacological research, aiding in studies focused on its stability and effects within clinical settings (Allibe et al., 2021).


Packaging

Packaged in poly bottles

Other Notes

may contain a slight excess of S

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of copper sulfide nanorod arrays on molecular templates.
Mao G, et al.
Nano Letters, 4(2), 249-252 (2004)
T P Mokone et al.
Journal of colloid and interface science, 351(1), 10-18 (2010-08-14)
Metal sulfide precipitation forms an important component of acid mine drainage remediation systems based on bacterial sulfate reduction. The precipitation reaction is thermodynamically favorable, but a number of technical issues remain. In this study the effect of metal to sulfide
Shusheng Zhang et al.
Analytical chemistry, 80(19), 7206-7212 (2008-09-02)
A novel and sensitive flow injection chemiluminescence assay for sequence-specific DNA detection based on signal amplification with nanoparticles (NPs) is reported in the present work. The "sandwich-type" DNA biosensor was fabricated with the thiol-functionalized capture DNA first immobilized on an
Vinod Kumar Gupta et al.
Journal of hazardous materials, 243, 179-186 (2012-11-06)
This study describes the effect of adsorption on methylene blue degradation using pectin-CuS nanocomposite (PCSNC). The nanocomposite was synthesized using co-precipitation methods followed by direct encapsulation with pectin. The synthesized nanocomposite was characterized by SEM, TEM, XRD, FTIR and UV-vis
Aaron E Saunders et al.
Nano letters, 6(12), 2959-2963 (2006-12-14)
The self-assembly of sterically stabilized colloidal copper sulfide nanodisks, 14-20 nm in diameter and 5-7 nm thick, was studied. The nanodisks were observed by electron microscopy and small-angle X-ray scattering to form columnar arrays when evaporated as thin films from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service