Skip to Content
Merck
All Photos(1)

Documents

237329

Sigma-Aldrich

1-Chloro-2,4-dinitrobenzene

≥99%

Synonym(s):

2,4-Dinitrochlorobenzene, CDNB, DNCB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)2
CAS Number:
Molecular Weight:
202.55
Beilstein:
613161
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

solid

expl. lim.

22 %

bp

315 °C (lit.)

mp

48-50 °C (lit.)

solubility

alcohol: very slightly soluble (cold)(lit.)
benzene: soluble(lit.)
carbon disulfide: soluble(lit.)
diethyl ether: soluble(lit.)
water: insoluble (practically)(lit.)

SMILES string

[O-][N+](=O)c1ccc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C6H3ClN2O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H

InChI key

VYZAHLCBVHPDDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chloro-2,4-dinitrobenzene on incubation with erythrocyte undergoes conjugation with erythrocyte glutathione (GSH) to form 2,4-dinitrophenyl-S-glutathione. It is an irreversible inhibitor of human thioredoxin reductase.

Application

1-Chloro-2,4-dinitrobenzene has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

381.2 °F - closed cup

Flash Point(C)

194 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y C Awasthi et al.
Blood, 58(4), 733-738 (1981-10-01)
Erythrocyte glutathione (GSH) can be rapidly depleted by incubating the cells with 1-chloro-2,4-dinitrobenzene (CDNB), which forms 2,4-dinitrophenyl-S-glutathione with GSH through the reaction catalyzed by glutathione S-transferase. GSH-CDNB conjugate thus formed stays undegraded within the erythrocytes. This indicates that in the
E S Arnér et al.
The Journal of biological chemistry, 270(8), 3479-3482 (1995-02-24)
Human thioredoxin reductase is a dimeric enzyme that catalyzes reduction of the disulfide in oxidized thioredoxin by a mechanism involving transfer of electrons from NADPH via FAD to a redox-active disulfide. 1-Chloro-2,4-dinitrobenzene (DNCB) is an alkylating agent used for depleting
Cynthia M F Monaco et al.
American journal of physiology. Endocrinology and metabolism, 318(1), E44-E51 (2019-12-04)
Sexual dimorphism in mitochondrial respiratory function has been reported in young women and men without diabetes, which may have important implications for exercise. The purpose of this study was to determine if sexual dimorphism exists in skeletal muscle mitochondrial bioenergetics
Boguen Kwon et al.
Nutrients, 13(3) (2021-04-04)
Cone of Pinus densiflora (CP), or Korean red pinecone, is a cluster of Pinus densiflora fruit. CP has also been verified in several studies to have anti-oxidation, anti-fungal, anti-bacterial, and anti-melanogenic effects. However, anti-inflammatory effects have not yet been confirmed
Jochem W van der Veen et al.
Archives of toxicology, 87(4), 763-766 (2012-11-13)
Several in vitro DNA microarray studies have shown the importance of nuclear factor (erythroid-derived 2)-like 2 (Nrf2) in skin sensitization. Nevertheless, the exact in vivo role of the Nrf2-Keap1 pathway during the induction of skin sensitization remains unknown. To study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service