Skip to Content
Merck
All Photos(3)

Documents

398047

Sigma-Aldrich

Resorcinol

ACS reagent, ≥99.0%

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

Assay

≥99.0%
99.0-100.5% (ACS specification)

form

powder and chunks

autoignition temp.

1126 °F

impurities

≤0.004 meq/g Titr. acid
≤0.005% insolubles

ign. residue

≤0.01%

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

solubility

H2O: 5%, clear, colorless (solution)

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Resorcinol (ACS reagent) can be used to determine nitrate concentrations in seawater using a spectrophotometric technique via nitration of resorcinol in acidified seawater.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic yolk-shell mesoporous silica microspheres with supported Au nanoparticles as recyclable high-performance nanocatalysts.
Yue Q, et al.
Journal of Material Chemistry A, 3(8), 4586-4594 (2015)
Dressler H.
Resorcinol: Its Uses and Derivatives, 5-12 (2013)
Mesomorphic properties of resorcinol.
Ossowska-Chrusciel MD, et al.
Journal of Molecular Structure, 1082, 103-113 (2015)
Size control and characterization of spherical carbon aerogel particles from resorcinol-formaldehyde resin.
Horikawa T, et al.
Carbon, 42(1), 169-175 (2004)
The chemistry of resorcinol carboxylation and its possible application to the CO2 removal from exhaust gases.
Barbarossa V, et al.
Journal of CO2 Utilization, 10, 50-59 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service