Skip to Content
Merck
All Photos(1)

Key Documents

467901

Supelco

Nickel(II) sulfate hexahydrate

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99.99% trace metals basis
grade:
for inorganic trace analysis
form:
solid

grade

for inorganic trace analysis

Quality Level

vapor density

2.07 (vs air)

Assay

≥99.99% trace metals basis

form

solid

impurities

≤0.002% N compounds
≤0.005% insolubles
<100 ppm total metallic impurities

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.005%
Co: ≤0.002%
Cu: ≤0.005%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
Mn: ≤0.002%
Na: ≤0.05%

SMILES string

O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nickel (II) sulfate hexahydrate (NiSO4·6H2O), commonly called retgersite, is a naturally occurring enantiomorphic mineral. It crystallizes in the monoclinic space group C2/c. Crystal structure of its various polymorphic forms have been investigated. The chiral nickel sulfate hexahydrate has been reported to induce asymmetric autocatalysis. The thermal degradation behavior of nickel(II) sulfate(VI) hexahydrate in air and helium atmosphere has been studied using thermogravimetry (TG) and X-ray powder diffraction (XRD).

Application


  • Improvement of ion chromatography with ultraviolet photometric detection and comparison with conductivity detection for the determination of serum cations.: This study explores advanced ion chromatography techniques for analyzing serum cations, where Nickel(II) sulfate hexahydrate may be used as a standard or in method development. The research compares ultraviolet photometric detection with conductivity detection, enhancing analytical methods for precise cation analysis in various samples, which can be crucial for environmental and clinical chemistry (Shintani H, 1985).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric induction by retgersite, nickel sulfate hexahydrate, in conjunction with asymmetric autocatalysis.
Matsumoto A, et al.
New. J. Chem., 39(9), 6742-6745 (2015)
Structure of monoclinic nickel (II) sulfate hexahydrate.
Gerkin RE and Reppart WJ.
Acta Crystallographica Section C, Crystal Structure Communications, 44(8), 1486-1488 (1988)
Polymorphism of nickel sulfate hexahydrate.
Angel RJ and Finger LW.
Acta Crystallographica Section C, Crystal Structure Communications, 44(11), 1869-1873 (1988)
Mechanism and kinetics of thermal decomposition of nickel (II) sulfate (VI) hexahydrate.
Tomaszewicz E and Kotfica M.
Journal of Thermal Analysis and Calorimetry, 77(1), 25-31 (2004)
Xiaohua Xu et al.
Particle and fibre toxicology, 9, 40-40 (2012-11-07)
It has been well recognized that toxicity of fine ambient air particulate matter (PM(2.5)) may depend on its chemical constituents, including components such as soluble metals that may theoretically exert distinctive effects. We have recently demonstrated an important effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service