Merck
  • Efficient [WO4](2-)-catalyzed chemical fixation of carbon dioxide with 2-aminobenzonitriles to quinazoline-2,4(1H,3H)-diones.
Efficient [WO4](2-)-catalyzed chemical fixation of carbon dioxide with 2-aminobenzonitriles to quinazoline-2,4(1H,3H)-diones.
Inorganic chemistry (2012-11-15)
Toshihiro Kimura, Hanako Sunaba, Keigo Kamata, Noritaka Mizuno
ABSTRACT
A simple monomeric tungstate, TBA(2)[WO(4)] (I, TBA = tetra-n-butylammonium), could act as an efficient homogeneous catalyst for chemical fixation of CO(2) with 2-aminobenzonitriles to quinazoline-2,4(1H,3H)-diones. Various kinds of structurally diverse 2-aminobenzonitriles could be converted into the corresponding quinazoline-2,4(1H,3H)-diones in high yields at atmospheric pressure of CO(2). Reactions of inactive 2-amino-4-chlorobenzonitrile and 2-amino-5-nitrobenzonitrile at 2 MPa of CO(2) also selectively proceeded. The present system was applicable to a g-scale reaction of 2-amino-5-fluorobenzonitrile (10 mmol scale) with CO(2) and 1.69 g of analytically pure quinazoline-2,4(1H,3H)-dione could be isolated. In this case, the turnover number reached up to 938 and the value was the highest among those reported for base-mediated systems so far. NMR spectroscopies showed formation of the corresponding carbamic acid through the simultaneous activation of both 2-aminobenzonitirile and CO(2) by I. Kinetic and computational studies revealed that I plays an important role in conversion of the carbamic acid into the product.
Product Description
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Supelco
Benzonitrile, suitable for HPLC, 99.9%
Sigma-Aldrich
Benzonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Tetrabutylammonium azide
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
2-Amino-4-chlorobenzonitrile, 99%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%