Skip to Content
Merck
All Photos(1)

Documents

359173

Sigma-Aldrich

Tetrabutylammonium nitrate

97%

Synonym(s):

N,N,N-Tributyl-1-butanaminium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(NO3)
CAS Number:
Molecular Weight:
304.47
Beilstein:
3921016
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

impurities

<3% tetrabutylammonium chloride

mp

116-118 °C (lit.)

SMILES string

[O-][N+]([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.NO3/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1(3)4/h5-16H2,1-4H3;/q+1;-1

InChI key

QHOKENWFMZXSEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Zhong et al.
Carbohydrate polymers, 94(1), 38-45 (2013-04-03)
In this article, a novel and high efficient solvent, tetra-n-Butylammonium Hydroxide (TBAH), was used for dissolution and isolation of straw cellulose from wheat straw. The composition analysis with gas chromatography (GC) and the spectroscopic characterization analysis conducted by X-Ray diffraction
Noelia Luque et al.
Journal of chromatography. A, 1248, 74-83 (2012-06-26)
The suitability of a mixed-mode sorbent made up of admicelles of sodium dodecyl sulphate (SDS) and tetrabutylammonium (TBA) to extract and preserve pesticides from river and underground water was assessed. Pesticides belonging to different structural groups (i.e. triazines, carbamates, phenylureas
Zhenfeng He et al.
Dalton transactions (Cambridge, England : 2003), 41(33), 10043-10051 (2012-05-29)
A new kind of organic-inorganic hybrid complexes based on polyoxometalate were synthesized through symmetrically grafting two adeninyl groups onto Anderson-type MnMo(6) clusters and encapsulating the clusters by organic surfactants. The resultant complexes exhibited thermal-induced dynamic self-assembly behaviors which greatly depended
David J Posson et al.
Nature structural & molecular biology, 20(2), 159-166 (2012-12-25)
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind
Toshihiro Kimura et al.
Inorganic chemistry, 51(23), 13001-13008 (2012-11-15)
A simple monomeric tungstate, TBA(2)[WO(4)] (I, TBA = tetra-n-butylammonium), could act as an efficient homogeneous catalyst for chemical fixation of CO(2) with 2-aminobenzonitriles to quinazoline-2,4(1H,3H)-diones. Various kinds of structurally diverse 2-aminobenzonitriles could be converted into the corresponding quinazoline-2,4(1H,3H)-diones in high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service