Skip to Content
Merck
All Photos(1)

Documents

72453

Sigma-Aldrich

Tetrabutylammonium bisulfate solution

~55% in H2O

Synonym(s):

Tetrabutylammonium hydrogen sulfate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
Beilstein:
3599663
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

~55% in H2O

SMILES string

OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

Application

Tetrabutylammonium bisulfate (Bu4NHSO4) can be used as a reagent along with potassium fluoride dihydrate to synthesize β-fluoro amine derivatives by the ring-opening reaction of aziridines. It may also be utilized as a catalyst in the hydrolysis of aziridines and epoxides to corresponding β-amino alcohols and 1,2-diols, respectively.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ren-Hua Fan et al.
The Journal of organic chemistry, 69(2), 335-338 (2004-01-17)
Potassium fluoride combined with tetrabutylammonium bisulfate is an efficient reagent to convert a variety of aziridines derived from cyclic and acyclic alkenes to beta-fluoro amine derivatives in high yield.
Ren-Hua Fan et al.
Organic & biomolecular chemistry, 1(9), 1565-1567 (2003-08-21)
Bu4NHSO4 (TBAHS) is an effective catalyst for the hydrolysis of aziridines and epoxides under mild and non-metal conditions to give the corresponding beta-amino alcohols and 1,2-diols in high yields. The catalyst can be recycled.
K M Frankowski et al.
Journal of dairy science, 97(9), 5356-5370 (2014-07-16)
Many food companies are trying to limit the amount of sodium in their products. Permeate, the liquid remaining after whey or milk is ultrafiltered, has been suggested as a salt substitute. The objective of this study was to determine the
Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of
David J Posson et al.
Nature structural & molecular biology, 20(2), 159-166 (2012-12-25)
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service