Skip to Content
Merck
All Photos(2)

Documents

86868

Sigma-Aldrich

Tetrabutylammonium bisulfate

puriss., ≥99.0% (T)

Synonym(s):

Tetrabutylammonium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
Beilstein:
3599663
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Quality Level

Assay

≥99.0% (T)

form

crystals

mp

169-173 °C

solubility

H2O: soluble 10 % (w/v), clear, colorless

SMILES string

OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium bisulfate, a quaternary ammonium salt, is a phase-transfer reagent.

Application

Tetrabutylammonium bisulfate may be employed in the following studies:
  • As a catalyst during the synthesis of β-amino alcohols and 1,2-diols.
  • As a phase transfer catalyst for the synthesis of asymmetrical ethers.
  • To compose the mobile phase for use in high performance liquid chromatographic (HPLC) analysis.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

348.1 °F - closed cup

Flash Point(C)

175.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erythrocytic adenosine monophosphate as an alternative purine source in Plasmodium falciparum.
Cassera MB, et al.
The Journal of Biological Chemistry, 283(47), 32889-32899 (2008)
Polyesters bearing furan moieties: IV. Solution and interfacial polycondensation of 2, 2'-bis (5-chloroformyl-2-furyl) propane with various diols and bisphenols.
Gharbi S, et al.
European Polymer Journal, 36(3), 463-472 (2000)
Phase-transfer synthesis of monoalkyl ethers of oligoethylene glycols.
Gibson T.
The Journal of Organic Chemistry, 45(6), 1095-1098 (1980)
Ren-Hua Fan et al.
Organic & biomolecular chemistry, 1(9), 1565-1567 (2003-08-21)
Bu4NHSO4 (TBAHS) is an effective catalyst for the hydrolysis of aziridines and epoxides under mild and non-metal conditions to give the corresponding beta-amino alcohols and 1,2-diols in high yields. The catalyst can be recycled.
K M Frankowski et al.
Journal of dairy science, 97(9), 5356-5370 (2014-07-16)
Many food companies are trying to limit the amount of sodium in their products. Permeate, the liquid remaining after whey or milk is ultrafiltered, has been suggested as a salt substitute. The objective of this study was to determine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service