Skip to Content
Merck
All Photos(1)

Key Documents

193119

Sigma-Aldrich

Tetrabutylammonium bromide

ReagentPlus®, ≥99.0%

Synonym(s):

TBAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(Br)
CAS Number:
Molecular Weight:
322.37
Beilstein:
3570983
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder, crystals or granules
solubility:
ethanol: 50 mg/mL, clear, colorless to light yellow

grade

reagent

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder, crystals or granules

pH

6.48 (30 °C)

mp

102-106 °C (lit.)

solubility

ethanol: 50 mg/mL, clear, colorless to light yellow

functional group

amine

SMILES string

[Br-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.BrH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

JRMUNVKIHCOMHV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium bromide is a quaternary ammonium compound widely used as a phase transfer catalyst. TBAB decreases the retention time and removes peak tailing by acting as an ion pair reagent during the chromatographic analysis of quaternary ammonium compounds. In the molten state, TBAB behaves like an ionic liquid, which is a promising green alternative to organic solvents in polymer synthesis. It participates as a catalyst during the solvent-free synthesis of biscoumarin and dihydropyrano[c]chromene derivatives.

Application

Tetrabutylammonium bromide may be used as a catalyst in the synthesis of following:
  • 1-cyano-2-thiophenylethane
  • 1-cyano-2-thiobutylethane
  • 1-cyano-2-(2-aminothiophenyl)ethane
  • methyl 3-thiophenylpropionate
  • methyl 3-thiobutylpropionate
  • methyl 2-methyl-3-thiophenylpropionate
  • methyl 2-methyl-3-thiobutylpropionate
  • diethyl 2-thiophenylsuccinate
  • diethyl 2-thiobutylsuccinate
  • 3-(4-chlorophenyl)-2-nitro-2-thiophenylpropane
  • 4-(4-chlorophenyl)-3-nitro-4-thiobutylbutane
  • 3-thiophenylcyclohexanone
  • 3-thioethylcyclohexanone
  • 4-methyl-4-thiophenylpentan-2-one
  • 4-methyl-4-thiobutylpentan-2-one
  • 4-thiophenylbutan-2-one
  • 4-thiobutylbutan-2-one
  • 3-thiophenylbutanal
  • 3-thiobutylbutanal

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel biobased polyurethanes synthesized from nontoxic phenolic diol containing l-tyrosine moiety under green media.
Mallakpour S, et al.
Journal of Polymers and the Environment, 18(4), 685-695 (2010)
Direct polyamidation in molten tetrabutylammonium bromide: novel and efficient green media.
Mallakpour S and Yousefian H.
Polym. Bull., 60(2-3), 191-198 (2008)
Tetrabutylammonium bromide (TBAB): a neutral and efficient catalyst for the synthesis of biscoumarin and 3, 4-dihydropyrano [c] chromene derivatives in water and solvent-free conditions.
Khurana JM and Kumar S.
Tetrahedron Letters, 50(28), 4125-4127 (2009)
Effect of analogue ions in normal-phase ion-pair chromatography of quaternary ammonium compounds.
Bluhm LH and Li T.
Journal of Chromatographic Science, 37(8), 273-276 (1999)
Interfacial chemistry behavior of phase transfer catalysis for hydroxide ion initiated reactions.
Xia L, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 317(1), 747-750 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service