Skip to Content
Merck
All Photos(1)

Key Documents

RAB0747

Sigma-Aldrich

Human TF / Serotransferrin ELISA Kit

Synonym(s):

Serotransferrin, TF, Transferrin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 ng/mL
standard curve range: 1.029-750 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TF(7018)

General description

Transferrin (TF) is an iron (Fe) transport protein and found in blood plasma and brain extracellular fluids. It belongs to the family of Fe-binding glycoproteins. TF is made of a single polypeptide chain with two domains and each domain consists of a Fe-binding site. It helps in the transport of Fe(III) between the cells in soluble and non-toxic forms. TF also acts as a bacteriostatic agent as well as a growth factor. It binds to the transferrin receptor to mediate the cellular uptake of bound Fe. The TF gene is mapped on the human chromosome at 3q22.1. The antibody pair provided in this kit recognizes human serotransferrin.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Schwarz et al.
Nutrients, 11(9) (2019-09-08)
Trace elements, like Cu, Zn, Fe, or Se, are important for the proper functioning of antioxidant enzymes. However, in excessive amounts, they can also act as pro-oxidants. Accordingly, trace elements influence redox-modulated signaling pathways, such as the Nrf2 pathway. Vice
Rosario Toro-Campos et al.
BMC pediatrics, 20(1), 225-225 (2020-05-20)
A central aim for pediatric nutrition is to develop infant formula compositionally closer to human milk. Milk fat globule membranes (MFGM) have shown to have functional components that are found in human milk, suggesting that addition of bovine sources of
Dahai Jiang et al.
Scientific reports, 10(1), 13334-13334 (2020-08-10)
Niemann-Pick C1 (NPC1) is a lysosomal cholesterol storage disorder, that severely affects the brain, and is caused by mutations in the NPC1 gene, which encodes an intracellular membrane transporter of non-esterified cholesterol. Therapeutic options for NPC1 are few, and classical
Akira Wada et al.
Journal of nutritional science and vitaminology, 64(1), 34-40 (2018-03-02)
In Japan, parenteral nutrition (PN) solutions are frequently administered to patients in the postoperative short-term period. In these cases, amino acid-containing peripheral parenteral nutrition (PPN) solutions, amino acid-free maintenance solutions or combinations of the two are used. However, consensus regarding
Halina Milnerowicz et al.
Inhalation toxicology, 30(13-14), 534-541 (2019-02-23)
Pregnancy and tobacco smoking (TS) each can cause increases in reactive oxygen species (ROS) production; this, in turn, can lead to disorders in iron management and disruption of the pro- and anti-oxidant balance. The aim of the study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service