Skip to Content
Merck
All Photos(2)

Key Documents

RAB0460

Sigma-Aldrich

Human TGF-β 1 ELISA Kit

Human TGF-β 1 ELISA Kit

for serum, plasma, and cell culture supernatants

Synonym(s):

TGF-beta-1, Transforming growth factor beta-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/mL
standard curve range: 0.0823-60 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TGFB1(7040)

General description

The Human TGF-β 1 ELISA (Enzyme-Linked mmunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human TGF-β 1 in serum, plasma, cell culture supernatants.

Immunogen

Recombinant Human TGF-β 1

Application

For research use only. Not for use in diagnostic procedures.
Human transforming growth factor-β1 (TGF-β1) ELISA Kit has been used to measure level of TGF-β1 released from tubular cells.[1]

Biochem/physiol Actions

Transforming growth factor-β1 (TGF-β1) is a pluripotential cytokine that plays a vital role in the homeostasis of peripheral CD4+CD25+ regulatory T cells and regulation of FOXP3 (forkhead box P3) expression in vivo. Binding of TGF-β1 to its receptor, activates members of the SMAD protein family involved in gene regulation.[2] Study shows that reduced expression of circulating transforming growth factor-β1 (TGF-β1) might contribute to systemic lupus erythematosus (SLE) development. Polymorphism of TGF-β1 is associated with pathogenesis of rheumatoid arthritis (RA) and pneumoconiosis.[3][4] Serum TGF-β1 expression is elevated in systemic sclerosis (SSc) patients.[5] Serum TGF-β1 functions as a potential biomarker of Guillain-Barré syndrome (GBS) and SSc.[5][6]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TGF-beta1 maintains suppressor function and Foxp3 expression in CD4+CD25+ regulatory T cells.
Marie JC
The Journal of Experimental Medicine, 201(7), 1061-1067 (2005)
Yali Zhou et al.
Experimental and therapeutic medicine, 21(6), 617-617 (2021-05-04)
The present study aimed to evaluate changes in bone mineral density, 25-hydroxyvitamin D3 [25-(OH)D3] and inflammatory factors in patients with hyperthyroidism, in order to determine the correlations with the pathogenesis of hyperthyroidism. A total of 55 patients with hyperthyroidism (observation
Heparanase is a key player in renal fibrosis by regulating TGF-? expression and activity.
Masola V
Biochimica et Biophysica Acta, 1843(9), 2122-2128 (2014)
XueXuan Zhang et al.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases, 59, 110-117 (2017-04-19)
To investigate PI3K-Akt-mTOR signaling pathway changes and the proliferation of FoxP3+Treg cells in patients with active tuberculosis. We isolated PBMCs and CD4+CD25+FoxP3+Treg cells from peripheral blood collected from patients with active tuberculosis and healthy controls. We compared the proportion and
Qiao Huang et al.
Experimental and therapeutic medicine, 18(1), 692-698 (2019-07-02)
NEF is a tumor suppressing long non-coding (lnc)RNA in hepatocellular carcinoma. Based on current literature, the involvement of NEF in other human diseases is still unknown. The current study aimed to investigate the potential involvement of NEF in glioma, which

Questions

Reviews

Active Filters

    • Review 1
    • Votes 0
    1 out of 5 stars.

    Do not recommend

    Does not come with the user guide. Tried to find it online and could not find it. It is so tedious.

    Helpful?

    1. Response from MilliporeSigma:

      The protocol for RAB0460 is listed on the COA and is consistent from lot to lot. We apologize the document was not easily accessible on our website and are making adjustments for easy access to a sample COA. In the meantime, please reach out to technical service for a copy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service