Skip to Content
Merck
All Photos(2)

Key Documents

RAB0402

Sigma-Aldrich

Human Platelet Factor-4 / CXCL4 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

C-X-C motif chemokine 4, Oncostatin-A, PF-4, Platelet factor 4, PF4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type urine
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 20.58-15000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PF4(5196)

General description

The Human PF-4 (Platelet Factor-4) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human PF-4 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human PF4

Application

For research use only. Not for use in diagnostic procedures.
Human Platelet Factor-4 (PF4)/ CXCL4 (chemokine (C-X-C motif) ligand 4) ELISA Kit has been used to measure PF-4 / CXCL4 serum level in chronic myelomonocytic leukemia (CMML) patients.[1]
Human Platelet Factor-4 / CXCL4 ELISA Kit has also been used in the measurement of platelet factor-4 in serum samples of prostate cancer[2][3] and pseudoxanthoma elasticum patients.[4]

Biochem/physiol Actions

Platelet factor-4 (PF-4) or chemokine (C-X-C motif) ligand 4 is chemotactic towards neutrophils and monocytes and has been shown to inhibit angiogenesis.[5] This protein associates with heparin and neutralizes it. It also neutralizes various other negatively charged proteoglycans. PF-4 / CXCL4 is implicated in cancer development.[6] PF-4 regulates the activity of proangiogenic growth factors. PF-4 / CXCL4 function as a biomarker for systemic sclerosis and early tumor growth.[7][8] PF-4 / CXCL4 is associated with various pathological conditions such as atherosclerosis, liver fibrosis and inflammation.[8]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Joseph et al.
Journal of thrombosis and haemostasis : JTH, 12(7), 1044-1053 (2014-04-29)
Heparin-induced thrombocytopenia (HIT) is an adverse immune-mediated response to unfractionated heparin and, less commonly, low molecular weight heparin. It is associated with a high thrombotic risk and the potential for limb and life-threatening complications. Argatroban is the only approved and
CXCL4-platelet factor 4, heparin-induced thrombocytopenia and cancer.
Sandset PM
Thrombosis Research, 129 Suppl 1, S97-100 (2012)
Kochawan Boonyawat et al.
Thrombosis research, 134(5), 957-962 (2014-09-11)
Heparin induced-thrombocytopenia (HIT) has been well recognized in Western countries. However, there are no data in the Thai population. We therefore investigated the prevalence of anti-platelet factor 4 (PF4)/heparin antibodies, HIT, and its thrombotic complications in Thai patients undergoing cardiac
Platelet-associated PF-4 as a biomarker of early tumor growth.
Cervi D
Blood, 111(3), 1201-1207 (2008)
Specific molecular signatures predict decitabine response in chronic myelomonocytic leukemia.
Meldi K
The Journal of Clinical Investigation, 125(5), 1857-1872 (2015)

Questions

  1. Could you please provide information on the detected value of the sample by RAB0402?

    1 answer
    1. From the testing of normal human plasma samples on the human PF-4 ELISA, levels were found to be around 500ng/ml.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service