Skip to Content
Merck
All Photos(1)

Key Documents

P38135

Sigma-Aldrich

Phosphate buffered saline

powder, pH 7.4, for preparing 5 L solutions

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

Product Name

Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions

Quality Level

form

powder

pH

7.4

solubility

water: soluble

application(s)

diagnostic assay manufacturing

Looking for similar products? Visit Product Comparison Guide

Application


  • Evaluation of anthelmintic efficacy of ethanolic leaf extract of Juglans regia L. on Ascaridia galli: a comprehensive in vitro and in vivo study.: This study explores the use of phosphate buffered saline as a diluent in testing the anthelmintic properties of Juglans regia leaf extract, providing valuable insights into its potential for parasitic control in veterinary applications (Mir et al., 2024).

  • Swelling kinetics of mixtures of soybean phosphatidylcholine and glycerol dioleate.: This study utilizes phosphate buffered saline to analyze the behavior of lipid mixtures, contributing to better understanding of biomaterials used in medical and cosmetic industries (Engstedt et al., 2024).

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in 5 liters of distilled or deionized water, will yield 0.01 M phosphate buffered saline (NaCl 0.138 M; KCl - 0.0027 M); pH 7.4, at 25 °C.

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung Y Cho et al.
Analytical and bioanalytical chemistry, 407(14), 4195-4206 (2015-03-31)
To help safeguard the food supply and detect the presence of undeclared food allergens and gluten, most producers and regulatory agencies rely on commercial test kits. Most of these are ELISAs with a few being PCR-based. These methods are very
Timothy J Green et al.
Fish & shellfish immunology, 47(1), 435-443 (2015-09-20)
The host-pathogen interactions between the Pacific oyster (Crassostrea gigas) and Ostreid herpesvirus type 1 (OsHV-1) are poorly characterised. Herpesviruses are a group of large, DNA viruses that are known to encode gene products that subvert their host's antiviral response. It
Mohidus Samad Khan et al.
Colloids and surfaces. B, Biointerfaces, 132, 264-270 (2015-06-09)
Viruses cause many infectious diseases and consequently epidemic health threats. Paper based diagnostics and filters can offer attractive options for detecting and deactivating pathogens. However, due to their infectious characteristics, virus detection using paper diagnostics is more challenging compared to
Beatriz del Rio et al.
Clinical and vaccine immunology : CVI, 15(9), 1429-1435 (2008-07-18)
Mucosal immunization is advantageous over other routes of antigen delivery because it can induce both mucosal and systemic immune responses. Our goal was to develop a mucosal delivery vehicle based on bacteria generally regarded as safe, such as Lactobacillus spp.
Yong N Li et al.
The Journal of comparative neurology, 516(5), 442-453 (2009-08-06)
Horizontal cells (HCs) are involved in establishing the center-surround receptive field organization of photoreceptor and bipolar cells. In many species, HCs respond differentially to colors and may play a role in color vision. An earlier study from our laboratory suggested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service