Skip to Content
Merck
All Photos(1)

Documents

P3813

Sigma-Aldrich

Phosphate buffered saline

powder, pH 7.4, for preparing 1 L solutions

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25

product name

Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions

form

powder

storage condition

dry at room temperature

color

white

pH

7.4

solubility

water: soluble

density

1.19 g/cm3 at 20—25 °C
1.22 g/cm3 at 20—25 °C

application(s)

diagnostic assay manufacturing
life science and biopharma
sample preparation

Looking for similar products? Visit Product Comparison Guide

General description

Phosphate-buffered saline (PBS) stands as a cornerstone in biological research due to its versatility and essential properties. One primary attribute is its isotonicity, closely resembling the pH, osmolarity, and ion concentrations of the human body. This makes PBS non-toxic to cells, playing a pivotal role in cell biology procedures such as washing cells, transporting tissues, and diluting cells and reagents. These applications ensure the preservation of cell viability and the maintenance of a physiological environment.

The buffering capacity of PBS, maintained through a phosphate-phosphoric acid equilibrium, provides a stable pH environment crucial for optimal enzyme activity and various biological processes. This makes PBS ideal for preparing reagents, protein dialysis, and immunohistochemistry, ensuring consistent and reliable results in experiments. Beyond cell biology, PBS may also find applications in diverse areas, including the creation of custom ELISA buffers to ensure optimal antibody-antigen interactions. Additionally, PBS can serve as a running buffer for Protein A/G chromatography, facilitating antibody purification based on specific antigen binding. In hematological and biochemical procedures, PBS is widely used for washing cells and tissues.

In summary, PBS emerges as an indispensable tool in both biological and biochemical research, offering isotonicity, buffering capacity, simplicity, and stability. Its diverse applications in cell biology, reagent preparation, protein purification, and various biological assays underscore its significance in advancing scientific knowledge and understanding.

Application

It has been utilized for rinsing fixed tissues and creating a suitable buffer environment in the immunolocalization process(1)It has been employed as a diluent to assess the performance of a ELISA test kit(2)It has been applied as a buffer in a immunohistochemistry procedure(3)
Phosphate buffered saline has been used:
  • in the washing solutions during immunofluorescence procedure.
  • for the suspension of bacterial cells.
  • during the fixation and immunostaining procedures.
  • for rinsing fixed tissues and creating a suitable buffer environment in the immunolocalization process
  • as a diluent to assess the performance of an ELISA test kit
  • as a buffer in an immunohistochemistry procedure

Features and Benefits

  • High-quality product suitable for multiple research applications
  • Can be used as a buffer component in biological and biochemical applications
  • Highly soluble in water

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in one liter of distilled or deionized water, will yield 0.01 M phosphate buffered saline (NaCl 0.138 M; KCl - 0.0027 M); pH 7.4, at 25 °C.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic changes in lymphocyte GRK2 levels in cardiac transplant patients: a biomarker for left ventricular function.
Bonita RE et al.
Clinical and Translational Science, 3, 14-14 (2010)
Jennifer L Matthews et al.
Zebrafish, 15(3), 279-290 (2018-01-26)
Sperm cryopreservation is a highly efficient method for preserving genetic resources. It extends the reproductive period of males and significantly reduces costs normally associated with maintenance of live animal colonies. However, previous zebrafish (Danio rerio) cryopreservation methods have produced variable
Development of an internal control for evaluation and standardization of a quantitative PCR assay for detection of Helicobacter pylori in drinking water.
Sen K et al.
Applied and Environmental Microbiology, 73, 7380-7380 (2007)
Effect of the microtubule polymerizing agent taxol on contraction, Ca2+ transient and L-type Ca2+ current in rat ventricular myocytes.
Howarth FC et al.
The Journal of Physiology, 516, 409-409 (1999)
Anirudh Sharma et al.
Scientific reports, 8(1), 4916-4916 (2018-03-22)
Citrate-stabilized iron oxide magnetic nanoparticles (MNPs) were coated with one of carboxymethyl dextran (CM-dextran), polyethylene glycol-polyethylene imine (PEG-PEI), methoxy-PEG-phosphate+rutin, or dextran. They were characterized for size, zeta potential, hysteresis heating in an alternating magnetic field, dynamic magnetic susceptibility, and examined

Articles

This is a protocol for a Membrane Integrity test for Lipid-PAMPA Artificial Membranes

Protocols

ICC Cell Capture Imaging Reagent simplifies imaging, ideal for rare cell and low cell count samples.

ICC Cell Capture Imaging Reagent simplifies imaging, ideal for rare cell and low cell count samples.

ICC Cell Capture Imaging Reagent simplifies imaging, ideal for rare cell and low cell count samples.

ICC Cell Capture Imaging Reagent simplifies imaging, ideal for rare cell and low cell count samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service