Merck
  • Oral immunization with recombinant lactobacillus plantarum induces a protective immune response in mice with Lyme disease.
Oral immunization with recombinant lactobacillus plantarum induces a protective immune response in mice with Lyme disease.
Clinical and vaccine immunology : CVI (2008-07-18)
Beatriz del Rio, Raymond J Dattwyler, Miguel Aroso, Vera Neves, Luciana Meirelles, Jos F M L Seegers, Maria Gomes-Solecki
ABSTRACT
Mucosal immunization is advantageous over other routes of antigen delivery because it can induce both mucosal and systemic immune responses. Our goal was to develop a mucosal delivery vehicle based on bacteria generally regarded as safe, such as Lactobacillus spp. In this study, we used the Lyme disease mouse model as a proof of concept. We demonstrate that an oral vaccine based on live recombinant Lactobacillus plantarum protects mice from tick-transmitted Borrelia burgdorferi infection. Our method of expressing vaccine antigens in L. plantarum induces both systemic and mucosal immunity after oral administration. This platform technology can be applied to design oral vaccine delivery vehicles against several microbial pathogens.
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, Autoclaved, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Phosphate buffered saline, BioUltra, washing buffer for peroxidase conjugates in Western Blotting, 10x concentrate
Sigma-Aldrich
Phosphate buffered saline, BioUltra, solution
Sigma-Aldrich
Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4
Sigma-Aldrich
Phosphate Buffered Saline, 10× PBS for Western blots and IP
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Phosphate Buffered Saline with 0.05% TWEEN® 20, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, pH 7.4, contains BSA, powder
Sigma-Aldrich
Phosphate buffered saline, pH 7.6 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet
Sigma-Aldrich
Phosphate buffered saline, with 5% nonfat milk, powder (dry milled), pH 7.3
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions