Skip to Content

System maintenance is scheduled for Sunday, October 11, 8:00 AM to 12:00 PM CST (Sunday, 11th October 2026, 03:00 PM to 07:00 PM CET). Quotes, price checks, and order status inquiries may be temporarily unavailable.

Merck

Key Documents

Skip To

08057

Phosphate buffered saline

tablet, pH 7.4 ( in solution), contains Tween® 20, BioUltra

Synonym(s):

PBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41105319
eCl@ss:
32129211
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate buffered saline, BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet

product line

BioUltra

Quality Segment

form

tablet

contains

TWEEN 20

color

colorless to white

pH

7.4 ( in solution)

solubility

water: 2 tablet/liter

UV absorption

λ: 260 nm Amax: ≤0.075, λ: 280 nm Amax: ≤0.050

Application

Phosphate buffered saline has been used in a study to determine that glioma immunotherapy with a combined vaccine prepared from tumor cells and endothelial cells might represent a new clinical strategy, as such combinations may theoretically affect both high-grade glioma cells and their environment. Phosphate buffered saline has also been used as a control injection in a study assessing place and direction learning by neonatal piglets.

Packaging

12 and 100 tablets in blister package

Preparation Note

1 tablet in 500 mL of water yields in 0.01 M phosphate buffer, 0.0027 M KCl, 0.14 M NaCl, 0.05% Tween, pH 7.4

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3813P4417P5368
description

BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet

description

powder, pH 7.4, for preparing 1 L solutions

description

tablet

description

BioPerformance Certified, pH 7.4

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

product line

BioUltra

product line

-

product line

-

product line

-

form

tablet

form

powder

form

tablet

form

powder

solubility

water: 2 tablet/liter

solubility

water: soluble

solubility

water: 1 tablet/200 mL, water: soluble

solubility

water: 9.07-10.0 g/L, clear, colorless

pH

7.4 ( in solution)

pH

7.4

pH

7.2-7.6 (1 tablet/200 mL), 7.4

pH

7.4

color

colorless to white

color

white

color

white

color

white


Still not finding the right product?

Explore all of our products under Phosphate buffered saline


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters