Skip to Content
Merck
All Photos(1)

Key Documents

M8042

Sigma-Aldrich

Minimum Essential Medium Eagle

Alpha Modification, with nucleosides and sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Minimum Essential Medium Eagle, Alpha Modification, with ribonucleosides, deoxyribonucleosides and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

Hanks’ salts (2% CO2): no
glucose: yes
sodium pyruvate: yes
phenol red: yes
HEPES: no
L-glutamine: no
stable glutamine: no
Earle’s salts (5% CO2): yes
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins. Often used in cytogenetic analysis when supplemented with fetal bovine serum (FBS).

Application

Minimum Essential Medium Eagle has been used for the isolation of mesenchymal stem cell, adipose-derived stem cell, and osteoblast.
Minimum Essential Medium Eagle is used for primary osteoblast cultures and for isolation of polymorphonuclear leukocytes (PMN) from umbilical cord blood.

Biochem/physiol Actions

MEM usually autoclaved with 0.5% bactopeptone at 15 psi for 20 min, cooled and then neutralized with NaHCO3, consistently supported good cell growth. A similar result was obtained when the MEM and bactopeptone were autoclaved separately. The cells grow, initially, as a monolayer and subsequently become a stationary suspension.

Reconstitution

Supplement with 0.292 g/L L-glutamine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A previously functional tetracycline-regulated transactivator fails to target gene expression to the bone
Schmidt E and Eriksson M
BMC Research Notes, 4(1), 282-282 (2011)
Eva Schmidt et al.
BMC research notes, 4, 282-282 (2011-08-13)
The tetracycline-controlled transactivator system is a powerful tool to control gene expression in vitro and to generate consistent and conditional transgenic in vivo model organisms. It has been widely used to study gene function and to explore pathological mechanisms involved
Isolation of adult stem cells and their differentiation to Schwann cells
Progenitor Cells, 47-57 (2012)
L Keay
Biotechnology and bioengineering, 19(3), 399-411 (1977-03-01)
The growth of L-60TM cells (a suspension culture adapted L-cell) on media composed of MEM (minimum essential medium (Eagle)) and bactopeptone autoclaved together or separately under a variety of conditions has veen determined. It has been found that MEM autoclaved
Janhavi Sharma et al.
Journal of immunological methods, 351(1-2), 30-35 (2009-10-06)
One of the initial steps in the inflammatory process involves the adherence and transmigration of circulating polymorphonuclear leukocytes (PMN) across the endothelial cell monolayer. One of the main constituents of the neutrophil phagosome that contributes to bacterial killing is myeloperoxidase

Articles

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service