Skip to Content
Merck
All Photos(1)

Documents

56416C

SAFC

Minimum Essential Medium

with Earle′s Balanced Salts, with 2.0 mM L-glutamine, with non-essential amino acids, without sodium bicarbonate, dry powder, suitable for cell culture

Pharma Manufacturing

Synonym(s):

Minimum Essential Medium Eagle, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

description

for research or for further manufacturing use

Quality Level

form

dry powder

technique(s)

cell culture | mammalian: suitable

components

phenol red: 10.620 mg/L
L-glutamine: 292 mg/L
NaHCO3: no
Earle’s salts (5% CO2): yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Quantity

Formulated to contain 9.6 grams of powder per liter of medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohan Chang et al.
Cell cycle (Georgetown, Tex.), 19(11), 1391-1403 (2020-04-15)
Long non-coding RNA (lncRNA) SOX2 overlapping transcript (SOX2OT) has been shown to play an oncogenic role in diverse cancers, generating eight transcript variants. SOX2 is located in the third intron of SOX2OT. However, the biological function of SOX2OT in cervical
Tatsuhiro Tsujimoto et al.
World journal of gastroenterology, 15(44), 5533-5540 (2009-11-26)
To investigate the possible use of the multiple cytokine production modulator, Y-40138, as a novel immunotherapy in the rat nonalcoholic steatohepatitis (NASH) model. We allocated 6-wk-old male F344 rats to choline-supplemented, L-amino acid-defined (CSAA) diet (control group), CSAA diet +
Wenxin Wu et al.
Virology, 396(2), 178-188 (2009-11-17)
We studied cytokine responses to influenza virus PR8 (H1N1) and Oklahoma/309/06 (OK/06, H3N2) in a novel human lung tissue model. Exposure of the model to influenza virus rapidly activated the mitogen-activated protein kinase signaling (MAPK) pathways ERK, p38 and JNK.
Robert Nason et al.
Journal of the Association for Research in Otolaryngology : JARO, 10(2), 151-160 (2009-01-16)
Osteoclasts are the only cells capable of carrying out bone resorption and therefore are responsible for the osteolysis seen in infectious diseases such as chronic otitis media and infected cholesteatoma. Pseudomonas aeruginosa is the most common organism isolated from these
Chia-Chyi Liu et al.
PloS one, 3(3), e1810-e1810 (2008-03-20)
This work investigated the replication kinetics of the four dengue virus serotypes (DEN-1 to DEN-4), including dengue virus type 4 (DEN-4) recovered from an infectious cDNA clone, in Vero cells and in MRC-5 cells grown on Cytodex 1 microcarriers. DEN-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service