Skip to Content
Merck
All Photos(2)

Key Documents

K1001

Sigma-Aldrich

β-Estradiol-6-one

Synonym(s):

1,3,5-Estratriene-3,17β-diol-6-one, 3,17β-Dihydroxy-1,3,5(10)-estratrien-6-one, 6-Ketoestradiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O3
CAS Number:
Molecular Weight:
286.37
Beilstein:
3208779
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98.00% (TLC)

form

powder

solubility

chloroform: methanol (1:1): 19.60-20.40 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@H]3[C@@H](CC(=O)c4cc(O)ccc34)[C@@H]1CC[C@@H]2O

InChI

1S/C18H22O3/c1-18-7-6-12-11-3-2-10(19)8-14(11)16(20)9-13(12)15(18)4-5-17(18)21/h2-3,8,12-13,15,17,19,21H,4-7,9H2,1H3/t12-,13-,15+,17+,18+/m1/s1

InChI key

ZHTDDOWJIRXOMA-YVEZLPLXSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Mikola et al.
Bioconjugate chemistry, 3(2), 182-186 (1992-03-01)
(Aminooxy)butyl- and -hexylamines and alcohols were synthesized by the Ing-Manske modification of the Gabriel synthesis. The aminooxy group of these heterobifunctional spacer reagents is a far more powerful nucleophile than the amino or hydroxy group because of the oxygen atom
Anna Bogucka-Kocka et al.
Journal of AOAC International, 94(1), 9-16 (2011-03-12)
Eighteen species belonging to the Carex genus were checked for the presence and the amount of eight phenolic acids (p-hydroxybenzoic, vanillic, caffeic, syringic, protocatechuic, p-coumaric, sinapic, and ferulic) by means of HPLC. Both the free and bonded phenolic acids were
M Adamczyk et al.
Steroids, 62(6), 462-467 (1997-06-01)
6-Oxoestradiol (2) was protected as its bis[(2-trimethylsilylethoxy)methyl] ether (4) and converted to the corresponding oxime (4). The oxime (4) on reduction with zinc in ethanol afforded the bis-SEM ether of 6-alpha-aminoestradiol (5) in 96% epimeric excess. Subsequently, 5 was hydrolyzed
N A Cridland et al.
The EMBO journal, 9(6), 1859-1866 (1990-06-01)
Photochemical excitation of a simple derivative of oestradiol using light in the UV-A range totally, permanently and selectively inactivates the oestrogen receptor protein present in a Xenopus liver extract without affecting its overall size. Inactivation of the binding site proceeds
M Numazawa et al.
Biochemical pharmacology, 47(4), 717-726 (1994-02-11)
Aromatase catalyzes the conversion of androst-4-ene-3,17-dione to estrogen through sequential oxygenations at the 19-methyl group. Androst-4-ene-3,6,17-trione (AT) is a suicide substrate of aromatase, and the mechanism of inactivation of aromatase has been postulated to involve enzymatic oxygenation at the 19-position.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service