Skip to Content
Merck
All Photos(3)

Documents

D9378

Sigma-Aldrich

3,4-Dihydroxy-D-phenylalanine

≥95% (TLC), powder, DOPA enantiomer

Synonym(s):

3-(3,4-Dihydroxyphenyl)-D-alanine, D-3-Hydroxytyrosine, D-DOPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO4
CAS Number:
Molecular Weight:
197.19
Beilstein:
2417637
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

product name

3,4-Dihydroxy-D-phenylalanine, powder, ≥95%

Assay

≥95%

form

powder

mp

276-278 °C (lit.)

solubility

1 M HCl: soluble
ethanol: insoluble

storage temp.

−20°C

SMILES string

N[C@H](Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m1/s1

InChI key

WTDRDQBEARUVNC-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Dihydroxy-D-phenylalanine has been used as a standard in high-performance liquid chromatography (HPLC) analysis of L-DOPA extracted from both germinated and raw seed flours of velvet bean.

Biochem/physiol Actions

3,4-Dihydroxy-D-phenylalanine (D-DOPA) is a DOPA enantiomer. It exhibits antimicrobial activity. D-DOPA serves as a precursor for dopamine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Lu et al.
Electrophoresis (2020-09-20)
This review summarizes recent developments (over the past decade) in the field of microfluidics-based solutions for enantiomeric separation and detection. The progress in various formats of microchip electrodriven separations, such as MCE, microchip electrochromatography, and multidimensional separation techniques, is discussed.
Tomoya Kawazoe et al.
Chemical record (New York, N.Y.), 7(5), 305-315 (2007-10-10)
The flavoprotein D-amino acid oxidase (DAO) degrades the gliotransmitter D-Ser, a potent activator of N-methyl-D-aspartate-type glutamate receptors. A body of evidence suggests that DAO, together with its activator, G72 protein, may play a key role in the pathophysiology of schizophrenia.
Mark H Burrell et al.
ACS chemical neuroscience, 6(11), 1802-1812 (2015-09-01)
Tonic dopamine (DA) levels influence the activity of dopaminergic neurons and the dynamics of fast dopaminergic transmission. Although carbon fiber microelectrodes and fast-scan cyclic voltammetry (FSCV) have been extensively used to quantify stimulus-induced release and uptake of DA in vivo
Philipp Hörmann et al.
Cell death discovery, 7(1), 151-151 (2021-07-07)
L-3,4-Dihydroxyphenylalanin (L-DOPA or levodopa) is currently the most used drug to treat symptoms of Parkinson's disease (PD). After crossing the blood-brain barrier, it is enzymatically converted to dopamine by neuronal cells and restores depleted endogenous neurotransmitter levels. L-DOPA is prone
Anthony Bertucci et al.
Bioorganic & medicinal chemistry, 18(6), 2300-2303 (2010-02-24)
The activity of the coral Stylophora pystillata secretory carbonic anhydrase STPCA has been tested in presence of amino acids and amines. All the investigated compounds showed a positive, activating effect on k(cat) and have been separated in weak (K(A) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service