Skip to Content
Merck

Skip To

D1507

L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride

≥98% (HPLC), solid,  L-DOPA precursor

Synonym(s):

Methyl L-DOPA hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13NO4 · HCl
CAS Number:
Molecular Weight:
247.68
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Form:
solid
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, solid

form

solid

Quality Level

color

white to off-white

storage temp.

−20°C

SMILES string

Cl[H].COC(=O)[C@@H](N)Cc1ccc(O)c(O)c1

InChI

1S/C10H13NO4.ClH/c1-15-10(14)7(11)4-6-2-3-8(12)9(13)5-6;/h2-3,5,7,12-13H,4,11H2,1H3;1H/t7-;/m0./s1

InChI key

WFGNJLMSYIJWII-FJXQXJEOSA-N

Application

L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride has been used:
  • in treating 6-hydroxydopamine induced lesions mice serotonin neurons[1]
  • in tyrosinase activity in B16F10 skin melanoma cells
  • to test its neuroprotective effect in mesencephalic neurons

Biochem/physiol Actions

L-3,4-Dihydroxyphenylalanine methyl ester is a precursor to L-DOPA that crosses the blood-brain barrier. Antiparkinsonian agent L-DOPA methyl ester elicits antitumor functionality in leukemia and melanoma.

Disclaimer

Light sensitive

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
UC168SML0091N5751
form

solid

form

solid

form

powder

form

powder

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

color

white to off-white

color

white to off-white

color

white to tan

color

white to off-white


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sodium tanshinone IIA silate increases melanin synthesis by activating the MAPK and PKA pathways and protects melanocytes from H2 O2-induced oxidative stress
Zhong H, et al.
Royal Society of Chemistry Advances, 9(33), 18747-18757 (2019)
Intranasal administration of the dopaminergic agonists L-DOPA, amphetamine, and cocaine increases dopamine activity in the neostriatum: a microdialysis study in the rat
De Souza Silva MA, et al.
Journal of Neurochemistry, 68(1), 233-239 (1997)
Fabrizio Stocchi et al.
Movement disorders : official journal of the Movement Disorder Society, 25(12), 1881-1887 (2010-07-30)
Melevodopa hydrochloride plus carbidopa in effervescent tablets (M/C) is a readily soluble antiparkinsonian tablet formulation. A total of 221 patients with Parkinson's disease and motor fluctuations entered a randomized, double-blind, double-dummy, controlled parallel group study, which compared the effectiveness of



Global Trade Item Number

SKUGTIN
D1507-500MG04061832081885
D1507-1G04061833560068
D1507-5G04061833560075

Questions

Reviews

No rating value

Active Filters