Merck
  • Carbonic anhydrase activators. The first activation study of a coral secretory isoform with amino acids and amines.
Carbonic anhydrase activators. The first activation study of a coral secretory isoform with amino acids and amines.
Bioorganic & medicinal chemistry (2010-02-24)
Anthony Bertucci, Didier Zoccola, Sylvie Tambutté, Daniela Vullo, Claudiu T Supuran
ABSTRACT
The activity of the coral Stylophora pystillata secretory carbonic anhydrase STPCA has been tested in presence of amino acids and amines. All the investigated compounds showed a positive, activating effect on k(cat) and have been separated in weak (K(A) in the range of 21-126 microM), medium (10.1-19 microM) and strong enzyme activators (K(A) of 0.18-3.21 microM). D-DOPA was found to be the best coral enzyme activator, with an activation constant K(A) of 0.18 microM. This enhancement of STPCA activity, as well as previous enzyme inhibition results, might now be tested on living organisms to better understand the role played by these enzymes in the coral calcification processes.
Product Description
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
SAFC
L-Histidine
Sigma-Aldrich
L-Tyrosine, FG
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland