Skip to Content
Merck
All Photos(1)

Documents

A0835

Sigma-Aldrich

N-Acetyl-Asp-Glu-Val-Asp-al

≥95%, powder

Synonym(s):

Ac-DEVD-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N4O11
CAS Number:
Molecular Weight:
502.47
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic

Quality Level

Assay

≥95%

form

powder

color

white

solubility

H2O: 1 mg/mL

storage temp.

−20°C

SMILES string

[H]C(=O)C(CC(O)=O)NC(=O)C(NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(C)=O)C(C)C

InChI

1S/C20H30N4O11/c1-9(2)17(20(35)22-11(8-25)6-15(29)30)24-18(33)12(4-5-14(27)28)23-19(34)13(7-16(31)32)21-10(3)26/h8-9,11-13,17H,4-7H2,1-3H3,(H,21,26)(H,22,35)(H,23,34)(H,24,33)(H,27,28)(H,29,30)(H,31,32)

InChI key

UMBVAPCONCILTL-UHFFFAOYSA-N

Gene Information

Amino Acid Sequence

NAc-Asp-Glu-Val-Asp-al

General description

N-Acetyl-Asp-Glu-Val-Asp-al (Ac-DEVD-CHO) is a tetrapeptide with N-terminal acetylation and C-terminal aldehyde conjugation.

Application

N-Acetyl-Asp-Glu-Val-Asp-al has been used as a caspase inhibitor in muscle samples, smooth muscle cells and Jurkat T-cells.

Biochem/physiol Actions

N-Acetyl-Asp-Glu-Val-Asp-al (Ac-DEVD-CHO) aids in the recovery of stunned myocardium by its caspase inhibition functionality.
Reversible inhibitor of IL-1β converting enzyme (ICE); inhibits poly(ADP-ribose) polymerase cleavage by apopain (caspase 3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carles Solà-Riera et al.
Scientific reports, 9(1), 834-834 (2019-01-31)
Orthohantaviruses, previously known as hantaviruses, are zoonotic viruses that can cause hantavirus pulmonary syndrome (HPS) and hemorrhagic fever with renal syndrome (HFRS) in humans. The HPS-causing Andes virus (ANDV) and the HFRS-causing Hantaan virus (HTNV) have anti-apoptotic effects. To investigate
Florian Guisier et al.
PloS one, 12(7), e0180576-e0180576 (2017-07-04)
Prediction of treatment outcome of non-small cell lung cancer (NSCLC) with EGFR inhibitors on the basis of the genetic analysis of the tumor can be incorrect in case of rare or complex mutations, bypass molecular activation pathways, or pharmacodynamic variations.
Yong-Bo Li et al.
Insect biochemistry and molecular biology, 79, 73-86 (2016-10-26)
Autophagy regulates cell survival (or cell death in several cases), whereas apoptosis regulates cell death. However, the relationship between autophagy and apoptosis and the regulative mechanism is unclear. We report that steroid hormone 20-hydroxyecdysone (20E) promotes switching from autophagy to
An-Cheng Huang et al.
Journal of ethnopharmacology, 194, 1043-1050 (2016-11-12)
Bufalin, a component of Chan Su (frog), has been shown to have biological activities including anti-tumor effects. Gefitinib has been used as an anti-cancer drug in lung cancer patients; however, some patients eventually become gefitinib resistant. In this study, we
Meili Fu et al.
Biochemical and biophysical research communications, 471(2), 267-273 (2016-01-17)
The aim of the present study is to investigate the potential anti-hepatocellular carcinoma (HCC) cell activity by 4SC-202, a novel class I HDAC inhibitor (HDACi). The associated signaling mechanisms were also analyzed. We showed that 4SC-202 treatment induced potent cytotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service