Skip to Content
Merck
All Photos(1)

Key Documents

A9968

Sigma-Aldrich

N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide

≥95%

Synonym(s):

Ac-IETD-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38N6O12
Molecular Weight:
638.62
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(C)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CC(O)=O)C(=O)Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C27H38N6O12/c1-5-13(2)22(28-15(4)35)26(42)30-18(10-11-20(36)37)24(40)32-23(14(3)34)27(43)31-19(12-21(38)39)25(41)29-16-6-8-17(9-7-16)33(44)45/h6-9,13-14,18-19,22-23,34H,5,10-12H2,1-4H3,(H,28,35)(H,29,41)(H,30,42)(H,31,43)(H,32,40)(H,36,37)(H,38,39)/t13-,14+,18-,19-,22-,23-/m0/s1

InChI key

FHURKTIGZAIQGR-BQCLNCLCSA-N

Amino Acid Sequence

NAc-Ile-Glu-Thr-Asp-pNA

Application

Chromogenic substrate for caspase 8 and granzyme B.

Biochem/physiol Actions

IETD is the sequence of amino acid residues 172-175 of procaspase 3, which is cleaved during apoptosis to produce the p12 subunit and a p20 peptide that is further cleaved to form the p17 subunit of mature caspase 3.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nafiseh Paydarnia et al.
Molecular biology reports, 46(3), 3129-3140 (2019-04-03)
As one of the most prevalent malignancies, breast cancer still remains a significant risk for public health. Common therapeutic strategies include invasive surgery, chemotherapy and anti-herceptin antibodies. Adverse effects, drug resistance and low efficacy of current therapies necessitates the emergence
Maria João Valente et al.
Archives of toxicology, 90(8), 1959-1973 (2015-12-18)
Synthetic cathinones have emerged in recreational drug markets as legal alternatives for classical amphetamines. Though currently banned in several countries, 3,4-methylenedioxypyrovalerone (MDPV) is one of the most commonly abused cathinone derivatives worldwide. We have recently reported the potential of MDPV
Huan Yu et al.
Insect science, 27(6), 1158-1172 (2019-12-04)
Apoptosis plays critical roles in multiple biological processes in multicellular organisms. Caspases are known as important participators and regulators of apoptosis. Here, four novel caspase genes of Spodoptera exigua were cloned and characterized, which were designated as SeCasp-1, SeCasp-6, SeCasp-7
Xiaosheng Qu et al.
Toxicon : official journal of the International Society on Toxinology, 110, 74-78 (2016-01-01)
The present work investigated the effects of the nematocysts venom (NV) from the Chrysaora helvola Brandt (C. helvola) jellyfish on the human nasopharyngeal carcinoma cell line, CNE-2. The medium lethal concentration (LC50), quantified by MTT assays, was 1.7 ± 0.53 μg/mL (n = 5). An atypical
Yan-Yih Goh et al.
Scientific reports, 6, 36868-36868 (2016-11-15)
Copper complexes with potent anti-tumor effect have been extensively developed. Most investigations of their modes of action focused on the biomolecular targets but not the signal transduction between target binding and cell death. We have previously shown that the cytotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service