Skip to Content
Merck
All Photos(3)

Key Documents

542075

Sigma-Aldrich

Lutetium(III) chloride hexahydrate

≥99.99% trace metals basis

Synonym(s):

Lutetium trichloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LuCl3 · 6H2O
CAS Number:
Molecular Weight:
389.42
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
form:
crystalline

Quality Level

Assay

≥99.99% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: lutetium

impurities

≤100 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.O.Cl[Lu](Cl)Cl

InChI

1S/3ClH.Lu.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

SJOQNHYDCUXYED-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Lutetium(III) chloride hexahydrate can be used:
  • As a starting material to prepare luminescent lutetium aluminum garnet for the preparation of optical ceramics.
  • To fabricate a modified glassy carbon electrode to detect sulfite in tap water.
  • As a precursor to prepare LuPO4 nanophosphors for bioimaging.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of heat-treatment temperature on the luminescent properties of Lu2O3:Eu film prepared by Pechini sol?gel method
Hai Guo, et al.
Applied Surface Science, 1, 245-250 (2005)
Low temperature synthesis and optical properties of the R2O3:Eu3+ nanophosphors (R3+: Y, Gd and Lu) using TMA complexes as precursors
I.G.N. Silva, et al.
Optical Materials, 40, 41-48 (2015)
LuPO4:Nd3+ nanophosphors for dual-mode deep tissue NIR-II luminescence/CT imaging
Xiaodan Wang, et al
Journal of Luminescence, 209, 420-426 (2019)
Electrochemical synthesis of lutetium (III) hexacyanoferrate/poly (taurine) modified glassy carbon electrode for the sensitive detection of sulfite in tap water
Journal of the Electrochemical Society, 165 , B469-B469 (2018)
Radiation-induced preparation of pure and Ce-doped lutetium aluminium garnet and its luminescent properties
Jan Barta, et al.
Journal of Materials Chemistry, 22, 16590-16597 (2012)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service